Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04894993_cn
          See other NCOA7 CNV Assays ›
          Gene Symbol
          NCOA7
          Assay Reference Genome
          Location

          Chr.6:125794610 on build GRCh38
          Cytoband
          Assay ID Hs04894993_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          135112

          Gene Name:

          nuclear receptor coactivator 7

          Gene Aliases:

          ERAP140, ESNA1, NCOA7-AS, Nbla00052, Nbla10993, TLDC4, dJ187J11.3

          Location:

          Chr.6:125781115-125932034 on Build GRCh38

          Assay Gene Location:

          Within Intron 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          NCOA7 NM_001122842.3 NP_001116314.1
          NM_001199619.2 NP_001186548.1
          NM_001199620.2 NP_001186549.1
          NM_001199621.2 NP_001186550.1
          NM_181782.5 NP_861447.3
          XM_005266822.5 XP_005266879.1
          XM_006715340.5 XP_006715403.1
          XM_017010270.2 XP_016865759.1
          XM_017010272.3 XP_016865761.1
          XM_017010273.3 XP_016865762.1
          XM_017010274.3 XP_016865763.1
          XM_024446331.2 XP_024302099.1
          XM_024446332.2 XP_024302100.1
          XM_047418206.1 XP_047274162.1
          XM_047418207.1 XP_047274163.1
          AB074156.1 BAE45731.1
          AB074157.1 BAE45732.1
          AF493978.1 AAM27392.1
          AK127512.1 BAG54516.1
          AK294558.1 BAH11810.1
          AK307395.1
          AL832621.2 CAD90002.2
          AL832628.2 CAD89948.1
          BC036461.1 AAH36461.1
          BC137094.1 AAI37095.1
          BC137095.1 AAI37096.1
          BX537385.1 CAD97627.1
          DA770225.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6602879 Chr.6:125735201 - 125820466 on Build GRCh38 Duplication NCOA7-AS1 NCOA7 HEY2-AS1 HEY2
          nsv950505 Chr.6:125772255 - 125803354 on Build GRCh38 Duplication NCOA7-AS1 NCOA7
          nsv6619060 Chr.6:125794286 - 125794957 on Build GRCh38 Deletion NCOA7

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          response to oxidative stress
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of cellular response to oxidative stress
          transcription coactivator activity
          protein binding
          ligand-dependent nuclear receptor binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline