Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Entrez Gene ID: | 2692 |
Gene Name: | growth hormone releasing hormone receptor |
Gene Aliases: |
GHRFR, GRFR, IGHD1B |
Location: |
Chr.7:30964021-30979531 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GHRHR | NM_000823.3 | NP_000814.2 | ||
XM_011515263.1 | XP_011513565.1 | |||
AF282259.1 | AAG15368.1 | |||
AF282260.1 | AAG15369.1 | |||
AF282261.1 | AAG15370.1 | |||
AF282262.1 | AAG15371.1 | |||
AY557192.1 | AAS59864.1 | |||
L01406.1 | AAA35890.1 | |||
L09237.1 | AAA58619.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv6316n100 | Chr.7:30609596 - 31145482 on Build GRCh38 | Gain | GHRHR GARS INMT-FAM188B AQP1 INMT ADCYAP1R1 FAM188B CRHR2 |
nsv527038 | Chr.7:30617851 - 31122331 on Build GRCh38 | Gain | GHRHR GARS INMT-FAM188B AQP1 INMT ADCYAP1R1 FAM188B CRHR2 |
esv2761316 | Chr.7:30615466 - 31127062 on Build GRCh38 | Gain | GHRHR GARS INMT-FAM188B AQP1 INMT ADCYAP1R1 FAM188B CRHR2 |
nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion | GHRHR INMT ZNRF2 CRHR2 NOD1 GARS ZNRF2P1 PDE1C PPP1R17 MIR550A1 FAM188B LOC401320 CCDC129 LOC101928268 NEUROD6 DKFZP586I1420 DPY19L1P1 AVL9 LOC100130673 INMT-FAM188B LINC01176 AQP1 LSM5 GGCT ADCYAP1R1 MIR550B1 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |