Search
Search

Entrez Gene ID: | 101929340 |
Gene Name: | uncharacterized LOC101929340 |
Gene Aliases: |
- |
Location: |
Chr.11:124759134-124765925 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929340 | NR_120577.1 | |||
| NR_120579.1 | ||||
| AK000493.1 | ||||
| CB997328.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556487 | Chr.11:124730862 - 124765248 on Build GRCh38 | Loss |
|