Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05322775_cn
          See other AP3S2 CNV Assays ›
          Gene Symbol
          AP3S2
          Assay Reference Genome
          Location

          Chr.15:89844805 on build GRCh38
          Cytoband
          Assay ID Hs05322775_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          10239

          Gene Name:

          adaptor related protein complex 3 subunit sigma 2

          Gene Aliases:

          AP3S3, sigma3b

          Location:

          Chr.15:89830599-89893994 on Build GRCh38

          Assay Gene Location:

          Within Intron 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          AP3S2 NM_005829.5 NP_005820.1
          NR_023361.2
          NR_037582.2
          AK222698.1 BAD96418.1
          AK294712.1 BAG57864.1
          AK312467.1 BAG35374.1
          AL110108.1
          BC002785.2 AAH02785.1
          BC007773.2 AAH07773.1
          BC010020.2 AAH10020.1
          DA777624.1
          DC340463.1
          X99459.1 CAA67824.1

          Target Gene Details

          Entrez Gene ID:

          100526783

          Gene Name:

          ARPIN-AP3S2 readthrough

          Gene Aliases:

          ARPIN, C15orf38, C15orf38-AP3S2

          Location:

          Chr.15:89830599-89912952 on Build GRCh38

          Assay Gene Location:

          Within Intron 8
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ARPIN-AP3S2 NM_001199058.2 NP_001185987.1
          AK000495.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv4242676 Chr.15:89843512 - 89852835 on Build GRCh38 Duplication AP3S2 ARPIN-AP3S2 MIR5094
          nsv5514328 Chr.15:89842808 - 89846684 on Build GRCh38 Deletion AP3S2 ARPIN-AP3S2
          nsv5522309 Chr.15:89842818 - 89846804 on Build GRCh38 Duplication AP3S2 ARPIN-AP3S2

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s)

          intracellular protein transport
          Golgi to vacuole transport
          anterograde axonal transport
          protein transport
          synaptic vesicle coating
          vesicle-mediated transport
          clathrin-coated vesicle cargo loading, AP-3-mediated
          synaptic vesicle recycling
          anterograde synaptic vesicle transport
          platelet dense granule organization
          melanosome assembly

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline