Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05337234_cn
          See other MCTP2 CNV Assays ›
          Gene Symbol
          MCTP2
          Assay Reference Genome
          Location

          Chr.15:94237297 on build GRCh38
          Cytoband
          Assay ID Hs05337234_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          55784

          Gene Name:

          multiple C2 and transmembrane domain containing 2

          Gene Aliases:

          -

          Location:

          Chr.15:94231366-94483952 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MCTP2 NM_001159643.2 NP_001153115.1
          NM_001385001.1 NP_001371930.1
          NM_001385002.1 NP_001371931.1
          NM_001385003.1 NP_001371932.1
          NM_001385004.1 NP_001371933.1
          NM_001385005.1 NP_001371934.1
          NM_001385006.1 NP_001371935.1
          NM_001385007.1 NP_001371936.1
          NM_001385008.1 NP_001371937.1
          NM_001385009.1 NP_001371938.1
          NM_001385010.1 NP_001371939.1
          NM_001385011.1 NP_001371940.1
          NM_018349.4 NP_060819.3
          NR_169526.1
          NR_169527.1
          NR_169528.1
          NR_169529.1
          NR_169530.1
          NR_169531.1
          NR_169532.1
          XM_011521774.3 XP_011520076.1
          XM_047432839.1 XP_047288795.1
          XM_047432840.1 XP_047288796.1
          XM_047432842.1 XP_047288798.1
          XM_047432843.1 XP_047288799.1
          AK090622.1
          AK300325.1 BAH13258.1
          AL832048.1
          AL832717.1
          BC041387.1 AAH41387.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv1052465 Chr.15:94011482 - 94804200 on Build GRCh38 Gain MCTP2 LINC01581 LOC105370987
          nsv6511648 Chr.15:94008168 - 94813217 on Build GRCh38 Duplication MCTP2 LINC01581 LOC105370987
          nsv1047585 Chr.15:93942818 - 94435921 on Build GRCh38 Gain MCTP2 LINC01581 LINC01580
          dgv463e214 Chr.15:94024364 - 94253291 on Build GRCh38 Gain MCTP2 LINC01581
          nsv5519560 Chr.15:94024533 - 94276083 on Build GRCh38 Duplication MCTP2 LINC01581
          esv3637305 Chr.15:94229922 - 94285926 on Build GRCh38 Loss MCTP2
          nsv833097 Chr.15:94162999 - 94351067 on Build GRCh38 Gain MCTP2
          nsv6514415 Chr.15:94234505 - 94285380 on Build GRCh38 Deletion MCTP2
          nsv4248958 Chr.15:94234496 - 94285380 on Build GRCh38 Deletion MCTP2
          dgv759e212 Chr.15:94236123 - 94285130 on Build GRCh38 Loss MCTP2
          dgv4730n54 Chr.15:94237012 - 94285237 on Build GRCh38 Loss MCTP2
          nsv523623 Chr.15:94233701 - 94285237 on Build GRCh38 Loss MCTP2
          esv2761821 Chr.15:94235856 - 94285033 on Build GRCh38 Loss MCTP2
          nsv6498463 Chr.15:94186016 - 94283892 on Build GRCh38 Deletion MCTP2
          nsv4531847 Chr.15:93698070 - 94477771 on Build GRCh38 Duplication MCTP2 LINC01581 LINC01580 LINC02207 LINC01579

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium-mediated signaling
          regulation of neurotransmitter secretion
          calcium ion binding
          calcium-dependent phospholipid binding
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline