Search
Search

Entrez Gene ID: | 10239 |
Gene Name: | adaptor related protein complex 3 subunit sigma 2 |
Gene Aliases: |
AP3S3, sigma3b |
Location: |
Chr.15:89830599-89893994 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP3S2 | NM_005829.5 | 6 | 4468 | NP_005820.1 |
| NR_023361.2 | 7 | 4587 | ||
| NR_037582.2 | 6 | 4464 | ||
| AL109702.1 | 1 | 796 | ||
| BC007773.2 | AAH07773.1 | |||
| DB322370.1 |
Entrez Gene ID: | 100526783 |
Gene Name: | ARPIN-AP3S2 readthrough |
Gene Aliases: |
ARPIN, C15orf38, C15orf38-AP3S2 |
Location: |
Chr.15:89830599-89912952 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPIN-AP3S2 | NM_001199058.2 | 10 | 5143 | NP_001185987.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1661 | Chr.15:89804782 - 89839712 on Build GRCh38 | Insertion |
|
Set Membership: |
|