Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05385921_cn
          See other PIGB CNV Assays ›
          Gene Symbol
          PIGB
          Assay Reference Genome
          Location

          Chr.15:55339563 on build GRCh38
          Cytoband
          Assay ID Hs05385921_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          9488

          Gene Name:

          phosphatidylinositol glycan anchor biosynthesis class B

          Gene Aliases:

          DEE80, EIEE80, GPI-MT-III, PIG-B

          Location:

          Chr.15:55319222-55355648 on Build GRCh38

          Assay Gene Location:

          Within Intron 9
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PIGB NM_004855.5 NP_004846.4
          XM_005254795.6 XP_005254852.1
          XM_011522235.4 XP_011520537.1
          XM_011522236.4 XP_011520538.1
          XM_011522237.3 XP_011520539.1
          XM_017022730.1 XP_016878219.1
          XM_017022731.2 XP_016878220.1
          XM_047433363.1 XP_047289319.1
          XM_047433364.1 XP_047289320.1
          XM_047433365.1 XP_047289321.1
          AK223330.1 BAD97050.1
          AK293666.1 BAG57112.1
          AK314706.1 BAG37253.1
          BC017711.1 AAH17711.1
          D42138.1 BAA07709.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2761899 Chr.15:53592144 - 55643724 on Build GRCh38 Gain+Loss RSL24D1 DNAAF4-CCPG1 PYGO1 RAB27A PIGB LOC124903496 MIR628 CCPG1 PIERCE2 UNC13C DNAAF4 WDR72 LOC105370829 PIGBOS1 PRTG
          nsv4242082 Chr.15:55003981 - 55380858 on Build GRCh38 Duplication RSL24D1 DNAAF4-CCPG1 RAB27A CCPG1 PIGB LOC105370829 MIR628 PIGBOS1
          nsv6515550 Chr.15:55306219 - 55474102 on Build GRCh38 Duplication DNAAF4-CCPG1 RAB27A CCPG1 PIERCE2 PIGB DNAAF4 MIR628 PIGBOS1
          nsv1043789 Chr.15:55108674 - 55800256 on Build GRCh38 Gain RSL24D1 DNAAF4-CCPG1 PYGO1 RAB27A CCPG1 PIERCE2 PIGB LOC124903496 DNAAF4 MIR628 PIGBOS1 PRTG
          nsv569523 Chr.15:55263404 - 55544506 on Build GRCh38 Gain DNAAF4-CCPG1 PYGO1 RAB27A CCPG1 PIERCE2 PIGB LOC124903496 DNAAF4 MIR628 PIGBOS1
          esv3569598 Chr.15:55270429 - 55410270 on Build GRCh38 Gain DNAAF4-CCPG1 RAB27A CCPG1 PIERCE2 PIGB MIR628 PIGBOS1
          nsv4247248 Chr.15:55307775 - 55493742 on Build GRCh38 Duplication DNAAF4-CCPG1 RAB27A CCPG1 PIERCE2 PIGB DNAAF4 MIR628 PIGBOS1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          GPI anchor biosynthetic process
          alpha-1,2-mannosyltransferase activity
          glycolipid mannosyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline