Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05462054_cn
          See other NQO1 CNV Assays ›
          Gene Symbol
          NQO1
          Assay Reference Genome
          Location

          Chr.16:69720770 on build GRCh38
          Cytoband
          Assay ID Hs05462054_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1728

          Gene Name:

          NAD(P)H quinone dehydrogenase 1

          Gene Aliases:

          DHQU, DIA4, DTD, NMOR1, NMORI, QR1

          Location:

          Chr.16:69709401-69726560 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          NQO1 NM_000903.3 NP_000894.1
          NM_001025433.2 NP_001020604.1
          NM_001025434.2 NP_001020605.1
          NM_001286137.2 NP_001273066.1
          AK223050.1 BAD96770.1
          AK297125.1 BAG59630.1
          AK297979.1 BAG60289.1
          AK298896.1 BAG61007.1
          AK307533.1
          AK309513.1
          AK312368.1 BAG35286.1
          AK316246.1 BAH14617.1
          BC007659.2 AAH07659.1
          BC107739.1 AAI07740.1
          BE903613.1
          CD014005.1
          DC331440.1
          J03934.1 AAA59940.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv4235244 Chr.16:69677025 - 69924863 on Build GRCh38 Duplication NFAT5 WWP2 NOB1 SNORD13H NQO1-DT NQO1 NQO1-AS1
          nsv517207 Chr.16:69711242 - 69738011 on Build GRCh38 Loss NQO1-DT NQO1
          nsv5532278 Chr.16:69677060 - 69924863 on Build GRCh38 Duplication NFAT5 WWP2 NOB1 SNORD13H NQO1-DT NQO1 NQO1-AS1
          nsv6515201 Chr.16:69713115 - 69724514 on Build GRCh38 Deletion NQO1
          esv3892878 Chr.16:69687512 - 69844258 on Build GRCh38 Gain NFAT5 WWP2 NOB1 SNORD13H NQO1-DT NQO1 NQO1-AS1
          esv3638928 Chr.16:69677077 - 69756070 on Build GRCh38 Gain NFAT5 NOB1 SNORD13H NQO1-DT NQO1 NQO1-AS1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          ubiquinone metabolic process
          xenobiotic metabolic process
          nitric oxide biosynthetic process
          response to oxidative stress
          synaptic transmission, cholinergic
          response to toxic substance
          removal of superoxide radicals
          protein catabolic process
          response to lipopolysaccharide
          cellular response to oxidative stress
          negative regulation of protein catabolic process
          vitamin E metabolic process
          vitamin K metabolic process
          innate immune response
          cell redox homeostasis
          response to caloric restriction
          negative regulation of ferroptosis
          RNA binding
          NAD(P)H dehydrogenase (quinone) activity
          cytochrome-b5 reductase activity, acting on NAD(P)H
          protein binding
          NADPH dehydrogenase (quinone) activity
          oxidoreductase activity
          identical protein binding
          NADH dehydrogenase (quinone) activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline