Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05511618_cn
          See other LRRC37A4P CNV Assays ›
          Gene Symbol
          LRRC37A4P
          Assay Reference Genome
          Location

          Chr.17:45516141 on build GRCh38
          Cytoband
          17q21.31
          Assay ID Hs05511618_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          55073

          Gene Name:

          leucine rich repeat containing 37 member A4, pseudogene

          Gene Aliases:

          LRRC37, LRRC37A4

          Location:

          Chr.17:45505883-45520523 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          LRRC37A4P NR_002940.2
          AK000982.1 BAA91454.1
          BC107850.1 AAI07851.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv9563 Chr.17:45494746 - 45629548 on Build GRCh38 Gain+Loss LRRC37A4P LOC644172 LOC105376839 MGC57346 MGC57346-CRHR1
          nsv1077843 Chr.17:45512133 - 45519934 on Build GRCh38 Duplication LRRC37A4P
          dgv927e212 Chr.17:45492771 - 45582619 on Build GRCh38 Loss LRRC37A4P
          dgv124e55 Chr.17:45464290 - 45589540 on Build GRCh38 Loss LRRC37A4P PLEKHM1 LOC105376839 MIR4315-1
          dgv928e212 Chr.17:45509002 - 45597851 on Build GRCh38 Gain LRRC37A4P LOC105376839
          nsv962329 Chr.17:45497931 - 45555048 on Build GRCh38 Duplication LRRC37A4P
          dgv5613n54 Chr.17:45513706 - 45519617 on Build GRCh38 Loss LRRC37A4P
          nsv442746 Chr.17:45508390 - 45571138 on Build GRCh38 Loss LRRC37A4P
          dgv3176n100 Chr.17:45496756 - 45633658 on Build GRCh38 Gain LRRC37A4P LOC644172 LOC105376839 MGC57346 MGC57346-CRHR1
          nsv509659 Chr.17:45481218 - 45534031 on Build GRCh38 Insertion LRRC37A4P PLEKHM1
          nsv575121 Chr.17:45514603 - 45524480 on Build GRCh38 Loss LRRC37A4P
          dgv5611n54 Chr.17:45511832 - 45520038 on Build GRCh38 Loss LRRC37A4P
          nsv161 Chr.17:45496487 - 45517762 on Build GRCh38 Insertion LRRC37A4P
          nsv953905 Chr.17:45497735 - 45533034 on Build GRCh38 Deletion LRRC37A4P
          dgv5610n54 Chr.17:45509805 - 45519155 on Build GRCh38 Loss LRRC37A4P
          esv26189 Chr.17:45496108 - 45634334 on Build GRCh38 Gain+Loss LRRC37A4P LOC644172 LOC105376839 MGC57346 MGC57346-CRHR1
          dgv123e55 Chr.17:45393394 - 45589540 on Build GRCh38 Loss LRRC37A4P PLEKHM1 ARHGAP27 LOC105376839 MIR4315-1
          nsv524234 Chr.17:45505316 - 45580555 on Build GRCh38 Loss LRRC37A4P
          esv33076 Chr.17:45493033 - 45598071 on Build GRCh38 Gain+Loss LRRC37A4P LOC105376839
          nsv428342 Chr.17:45366188 - 45573718 on Build GRCh38 Gain LRRC37A4P PLEKHM1 ARHGAP27 MIR4315-1
          dgv1760e59 Chr.17:45511903 - 45520201 on Build GRCh38 Duplication LRRC37A4P
          esv2762433 Chr.17:45496768 - 46710828 on Build GRCh38 Gain+Loss LOC107987253 NSFP1 STH MAPT LOC107987254 LOC644172 KANSL1 MGC57346 MGC57346-CRHR1 SPPL2C MAPT-IT1 CRHR1-IT1 MAPT-AS1 LRRC37A2 LRRC37A4P ARL17B LRRC37A CRHR1 ARL17A NSF LOC105376839 KANSL1-AS1
          nsv514848 Chr.17:45514695 - 45520131 on Build GRCh38 Gain LRRC37A4P
          esv2758691 Chr.17:45380520 - 45738657 on Build GRCh38 Gain+Loss CRHR1-IT1 LRRC37A4P PLEKHM1 LOC644172 ARHGAP27 LOC105376839 MGC57346 MGC57346-CRHR1 MIR4315-1
          dgv1473n106 Chr.17:45512034 - 45520134 on Build GRCh38 Duplication LRRC37A4P
          nsv820961 Chr.17:45512035 - 45520289 on Build GRCh38 Duplication LRRC37A4P

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0