Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05527855_cn
          See other KRTAP22-2 CNV Assays ›
          Gene Symbol
          KRTAP22-2
          Assay Reference Genome
          Location

          Chr.21:30590382 on build GRCh38
          Cytoband
          Assay ID Hs05527855_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          100288287

          Gene Name:

          keratin associated protein 22-2

          Gene Aliases:

          KAP22.2

          Location:

          Chr.21:30590105-30590397 on Build GRCh38

          Assay Gene Location:

          Overlaps - Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          KRTAP22-2 NM_001164434.1 NP_001157906.1
          AB096950.1 BAE46365.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv6546600 Chr.21:29796823 - 30635785 on Build GRCh38 Duplication CLDN8 KRTAP20-2 LINC00307 GRIK1 KRTAP24-1 KRTAP27-1 KRTAP13-1 KRTAP15-1 KRTAP6-3 KRTAP26-1 KRTAP20-4 KRTAP22-1 KRTAP19-7 CLDN17 MIR4327 KRTAP13-4 KRTAP19-3 KRTAP23-1 KRTAP25-1 KRTAP19-2 KRTAP20-1 KRTAP13-3 KRTAP19-1 KRTAP6-2 KRTAP19-5 KRTAP19-4 KRTAP6-1 KRTAP13-2 KRTAP19-6 KRTAP22-2
          nsv4290744 Chr.21:30551347 - 30621373 on Build GRCh38 Duplication KRTAP20-1 KRTAP6-2 KRTAP6-1 KRTAP6-3 KRTAP20-4 KRTAP22-1 KRTAP19-7 KRTAP22-2
          nsv4287786 Chr.21:30513966 - 30688897 on Build GRCh38 Deletion KRTAP20-1 KRTAP6-2 KRTAP20-2 KRTAP6-1 KRTAP6-3 KRTAP20-3 KRTAP20-4 KRTAP19-6 KRTAP22-1 KRTAP19-7 KRTAP22-2
          nsv5539790 Chr.21:5227438 - 39583816 on Build GRCh38 Insertion KRTAP21-3 LINC00189 MAP3K7CL LINC00307 LINC01683 HLCS LOC102724594 BAGE2 EPCIP KRTAP24-1 LINC03147 KRTAP13-1 SH3BGR KRTAP20-3 KRTAP26-1 LINC00310 ANKRD20A11P BTG3 LINC02573 TPTE CLDN17 URB1 POTED LINC01695 ADAMTS5 SOD1-DT CLIC6 BRWD1-AS2 LOC105369306 RNA5-8SN2 KRTAP19-5 LOC101930100 KRTAP19-4 RWDD2B MIR8069-1 LOC102724843 RNA18SN3 DSCR10 MIR4759 RNA28SN2 LINC01700 MIR6724-1 MIR6501 LOC102724560 ANKRD30BP2 LINC02943 LOC101928126 LTN1 KCNJ15 CHAF1B TTC3 CLDN14 LINC01548 LOC106780825 MIR6724-3 LOC102724354 KRTAP15-1 MIR125B2 LOC105369308 RNA5-8SN3 RBM11 LINC00317 D21S2088E OLIG1 MIR155 HEMK2 LINC03104 MRAP-AS1 KRTAP19-3 LINC01687 RNA45SN3 CCT8 LOC100506403 LINC01426 DOP1B LINC00158 LOC107987288 HMGN1 NCAM2 MORC3 KRTAP13-2 CH507-42P11.6 IL10RB LOC124905049 MIR548XHG HUNK MIR99AHG SOD1 KCNE2 KRTAP19-8 CLDN14-AS1 CLDN8 IFNGR2 DSCR4 SYNJ1 LINC00308 RNA5-8SN1 MIR6724-2 RIPPLY3 C21orf140 LOC101927745 LOC105372753 ETS2-AS1 LINC00114 GRIK1-AS3 LINC00113 LOC101927345 KRTAP8-1 MIR6130 KRTAP22-1 DONSON TMPRSS15 IFNAR2-IL10RB LOC105379504 LCA5L KRTAP23-1 SNORD74B PAXBP1 GRIK1-AS1 LOC102724334 LINC01690 LOC105379499 LINC00161 CYYR1-AS1 SCAF4 GET1-SH3BGR TTC3-AS1 SETD4-AS1 LINC01436 DNAJC28 LINC00649 ADAMTS1 MEMO1P1 USP25 LOC102723360 LOC107983987 LINC00515 LINC02920 MIR10396B RUNX1-IT1 LOC101928107 RNA18SN2 LOC105379514 LINC01423 LOC102724652 CYP4F29P MIR4327 RNA45SN1 SON LIPI KRTAP20-1 KRTAP13-3 VPS26C KRTAP6-2 CXADR TCP10L LINC00314 GET1 ATP5PO LINC01697 LINC01673 CRYZL1 LINC01689 LOC105372793 HLCS-AS1 PIGP LINC01667 MIR6508 MIR802 SNORA80A CBR1-AS1 KRTAP27-1 LOC110091777 ASMER1 BRWD1 MIRLET7C C21orf91-OT1 LOC102723475 ATP5PF LOC105372791 KRTAP7-1 NRIP1 RNA28SN3 KRTAP25-1 LOC124905050 KRTAP21-2 KRTAP19-2 KRTAP11-1 LOC105372762 LINC01549 CFAP298-TCP10L BRWD1-AS1 MIR155HG KCNJ6 CBR1 MIR548X ETS2 RNA28SN1 CFAP298 HSPA13 KRTAP20-2 KCNE1 MIS18A URB1-AS1 TIAM1-AS1 LOC102723553 BTG3-AS1 TMEM50B LINC01684 RUNX1-AS1 RPL23P2 KRTAP6-3 KRTAP20-4 SAMSN1 GRIK1-AS2 CBR3-AS1 APP-DT ITSN1 IFNAR1 LOC101928269 EPCIP-AS1 LINC00160 DYRK1A LINC00159 SMIM11 GABPA MIR3156-3 SETD4 NCOR1P4 DSCR8 KRTAP22-2 SAMSN1-AS1 CHODL MIR10396A GART PPP1R2P2 LINC01692 GRIK1 ABCC13 TIAM1 IL10RB-DT EVA1C C21orf91 MIR8069 ERG APP KRTAP13-4 LINC03105 RUNX1 PSMG1 LOC102723451 CBR3 LOC105377134 SMIM34 LOC101928398 MIR99A LOC102724428 KRTAP19-6 MIR3648-2 LINC01666 SLC5A3 LOC100288590 RNA18SN1 LINC01674 CYYR1 IGHV1OR21-1 SIM2 RNA45SN2 RCAN1 MIR3118-1 PAXBP1-AS1 TEKT4P2 MIR6724-4 OLIG2 LINC00945 KRTAP19-7 JAM2 BACH1-IT3 MRAP MIR3648-1 BACH1-IT2 USP16 BACH1 KRTAP21-1 IFNAR2 MRPS6 MRPL39 KRTAP19-1 LOC110091776 LINC01425 KRTAP6-1 DSCR9 LINC00320 KCNJ6-AS1 MIS18A-AS1
          esv2762105 Chr.21:30547730 - 30624847 on Build GRCh38 Gain KRTAP20-1 KRTAP6-2 KRTAP6-1 KRTAP6-3 KRTAP20-4 KRTAP22-1 KRTAP19-7 KRTAP22-2
          nsv1057467 Chr.21:30318003 - 30993921 on Build GRCh38 Gain KRTAP21-3 KRTAP20-2 KRTAP27-1 KRTAP13-1 KRTAP15-1 KRTAP6-3 KRTAP20-3 KRTAP26-1 KRTAP8-1 KRTAP20-4 KRTAP22-1 KRTAP19-7 KRTAP7-1 MIR4327 KRTAP13-4 KRTAP19-3 KRTAP23-1 KRTAP21-1 KRTAP21-2 KRTAP19-2 KRTAP20-1 KRTAP13-3 KRTAP11-1 KRTAP19-1 KRTAP6-2 KRTAP19-5 KRTAP19-4 KRTAP6-1 KRTAP13-2 KRTAP19-6 KRTAP22-2
          nsv587354 Chr.21:29978351 - 30620545 on Build GRCh38 Gain CLDN8 LINC00307 KRTAP24-1 KRTAP27-1 KRTAP13-1 KRTAP15-1 KRTAP6-3 KRTAP26-1 KRTAP22-1 KRTAP19-7 CLDN17 MIR4327 KRTAP13-4 KRTAP19-3 KRTAP23-1 KRTAP25-1 KRTAP19-2 KRTAP20-1 KRTAP13-3 KRTAP19-1 KRTAP6-2 KRTAP19-5 KRTAP19-4 KRTAP6-1 KRTAP13-2 KRTAP19-6 KRTAP22-2

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline