Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05537310_cn
          See other RUNX1 CNV Assays ›
          Gene Symbol
          RUNX1
          Assay Reference Genome
          Location

          Chr.21:34807905 on build GRCh38
          Cytoband
          Assay ID Hs05537310_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          861

          Gene Name:

          RUNX family transcription factor 1

          Gene Aliases:

          AML1, AML1-EVI-1, AMLCR1, CBF2alpha, CBFA2, EVI-1, PEBP2aB, PEBP2alpha

          Location:

          Chr.21:34787801-35049302 on Build GRCh38

          Assay Gene Location:

          Within Intron 8
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          RUNX1 NM_001001890.3 NP_001001890.1
          NM_001754.5 NP_001745.2
          XM_005261068.4 XP_005261125.1
          XM_005261069.5 XP_005261126.1
          XM_011529766.3 XP_011528068.1
          XM_011529767.3 XP_011528069.1
          XM_011529768.3 XP_011528070.1
          XM_047441007.1 XP_047296963.1
          XM_047441009.1 XP_047296965.1
          XM_047441010.1 XP_047296966.1
          XM_047441011.1 XP_047296967.1
          XM_047441012.1 XP_047296968.1
          XM_047441013.1 XP_047296969.1
          XM_047441014.1 XP_047296970.1
          XM_047441015.1 XP_047296971.1
          AK226159.1
          BC110828.1 AAI10829.1
          BC136380.1 AAI36381.1
          BC136381.1 AAI36382.1
          BC144053.1 AAI44054.1
          D43968.1 BAA07903.1
          D43969.1 BAA07904.1
          D89788.1 BAA14020.1
          D89789.1 BAA14021.1
          D89790.1 BAA14022.1
          L34598.1 AAA51720.1
          S60998.1
          S76350.1 AAB33731.1
          X79549.1 CAA56092.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv5550749 Chr.21:34567808 - 34891808 on Build GRCh38 Duplication RUNX1 LINC01426 LINC00160 CLIC6 RUNX1-AS1 RCAN1
          nsv5539790 Chr.21:5227438 - 39583816 on Build GRCh38 Insertion MAP3K7CL DSCR9 LOC105379514 APP TMEM50B LINC00161 C21orf91 MIR99A MIR548XHG KRTAP22-1 LOC105372762 PPP1R2P2 MEMO1P1 LOC105372753 CLDN17 CFAP298-TCP10L GRIK1-AS3 LOC105369308 LINC01423 RNA18SN3 ETS2-AS1 CRYZL1 KRTAP20-4 TMPRSS15 KRTAP6-3 VPS26C TTC3 RNA28SN1 KCNE2 LOC105379504 LINC01666 KRTAP21-2 RNA18SN1 ANKRD30BP2 ERG SON CHODL GET1-SH3BGR CLDN14 KRTAP8-1 ANKRD20A11P KRTAP6-2 SH3BGR LOC102724354 RNA45SN3 LOC110091777 MIR6724-4 LOC124905050 LINC01436 LOC107987288 KRTAP11-1 MIR802 LINC00515 RNA5-8SN3 RUNX1 LINC00314 BACH1-IT2 CBR1 LOC102724843 MIR3156-3 LINC00317 MRAP-AS1 LINC01695 OLIG2 MIS18A KCNE1 KCNJ6-AS1 TTC3-AS1 KRTAP7-1 C21orf140 CLIC6 IFNAR2-IL10RB KRTAP19-4 KRTAP26-1 LINC02920 KRTAP21-1 LINC00160 IFNAR1 BACH1-IT3 PSMG1 NCAM2 LINC01692 LINC02943 GRIK1-AS1 LINC01687 LOC101928107 LINC01700 MIR6501 LINC00307 BTG3 SAMSN1 LOC102724652 MIR3648-2 KRTAP6-1 IGHV1OR21-1 OLIG1 KRTAP19-6 MIRLET7C MIR6724-2 CBR3 SOD1 USP25 RUNX1-AS1 RNA45SN1 USP16 LINC01673 KRTAP19-7 RNA5-8SN2 LOC102723475 EVA1C BRWD1-AS2 KRTAP13-3 LINC01697 MIR4327 LOC102723553 LTN1 SIM2 LINC01684 RCAN1 GABPA GRIK1-AS2 MIR6724-3 TEKT4P2 HEMK2 ADAMTS5 IFNGR2 LOC101930100 LINC01683 LINC01689 ITSN1 CH507-42P11.6 KRTAP13-4 KRTAP25-1 RNA45SN2 LOC101928269 ADAMTS1 LOC124905049 SMIM11 KRTAP20-2 MIR4759 SETD4 POTED C21orf91-OT1 CYP4F29P MIR8069 LOC105372793 DSCR8 KRTAP22-2 CXADR URB1 SLC5A3 LINC00158 SOD1-DT LOC100506403 RIPPLY3 DYRK1A RNA28SN2 SMIM34 CFAP298 KCNJ6 LOC102723451 ETS2 CYYR1-AS1 MRAP LINC03104 NCOR1P4 DSCR10 MIR10396A DSCR4 CCT8 BACH1 LINC00320 GART RUNX1-IT1 MORC3 MIR6130 BRWD1-AS1 PAXBP1 MIS18A-AS1 KRTAP19-3 CBR3-AS1 SETD4-AS1 LOC102724560 MIR10396B RNA5-8SN1 LINC01690 MIR3118-1 MRPL39 ATP5PO HLCS-AS1 EPCIP-AS1 MIR6724-1 IL10RB-DT LINC00310 MIR125B2 SAMSN1-AS1 LINC00189 LINC00114 BAGE2 SYNJ1 GET1 D21S2088E RNA18SN2 PAXBP1-AS1 KRTAP19-5 CLDN8 LINC00308 LOC101928398 HMGN1 KRTAP19-2 LINC00159 LOC101927745 RBM11 PIGP MIR155HG KRTAP13-2 CYYR1 LOC102723360 TIAM1-AS1 KRTAP13-1 SNORD74B DONSON MIR548X IL10RB KRTAP19-8 KRTAP19-1 LINC01549 LOC105372791 MRPS6 LINC00113 LOC106780825 TPTE SNORA80A CBR1-AS1 TIAM1 APP-DT LINC01674 TCP10L LOC102724594 JAM2 EPCIP HUNK LINC01667 ASMER1 ATP5PF MIR8069-1 CLDN14-AS1 DOP1B MIR99AHG RNA28SN3 MIR3648-1 RWDD2B KRTAP21-3 LCA5L SCAF4 BRWD1 LINC01548 KRTAP15-1 LINC01425 IFNAR2 KRTAP20-3 LOC107983987 KRTAP23-1 NRIP1 LINC00945 MIR155 LINC00649 MIR6508 LOC105377134 LINC03147 KRTAP24-1 HSPA13 LOC105379499 ABCC13 LOC105369306 LOC110091776 KRTAP20-1 URB1-AS1 LOC101927345 LOC102724428 BTG3-AS1 KCNJ15 CHAF1B LINC02573 LINC01426 LIPI LOC101928126 HLCS RPL23P2 DNAJC28 LOC102724334 GRIK1 LOC100288590 LINC03105 KRTAP27-1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ossification
          chondrocyte differentiation
          myeloid leukocyte differentiation
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          regulation of plasminogen activation
          hemopoiesis
          myeloid cell differentiation
          neuron differentiation
          negative regulation of granulocyte differentiation
          positive regulation of granulocyte differentiation
          positive regulation of interleukin-2 production
          positive regulation of collagen biosynthetic process
          negative regulation of CD4-positive, alpha-beta T cell differentiation
          positive regulation of CD8-positive, alpha-beta T cell differentiation
          regulation of cell differentiation
          positive regulation of angiogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          peripheral nervous system neuron development
          positive regulation of developmental process
          positive regulation of multicellular organismal process
          regulation of cardiac muscle cell proliferation
          cardiac muscle tissue regeneration
          hematopoietic stem cell proliferation
          positive regulation of extracellular matrix organization
          regulation of connective tissue replacement
          regulation of multicellular organismal development
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcription corepressor binding
          transcription coactivator binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          calcium ion binding
          protein binding
          ATP binding
          protein homodimerization activity
          protein heterodimerization activity
          DNA-binding transcription factor binding
          sequence-specific double-stranded DNA binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline