Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05551226_cn
          See other RUNX1 CNV Assays ›
          Gene Symbol
          RUNX1
          Assay Reference Genome
          Location

          Chr.21:34797244 on build GRCh38
          Cytoband
          Assay ID Hs05551226_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          861

          Gene Name:

          RUNX family transcription factor 1

          Gene Aliases:

          AML1, AML1-EVI-1, AMLCR1, CBF2alpha, CBFA2, EVI-1, PEBP2aB, PEBP2alpha

          Location:

          Chr.21:34787801-35049302 on Build GRCh38

          Assay Gene Location:

          Within Intron 9
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          RUNX1 NM_001001890.3 NP_001001890.1
          NM_001754.5 NP_001745.2
          XM_005261068.4 XP_005261125.1
          XM_005261069.5 XP_005261126.1
          XM_011529766.3 XP_011528068.1
          XM_011529767.3 XP_011528069.1
          XM_011529768.3 XP_011528070.1
          XM_047441007.1 XP_047296963.1
          XM_047441009.1 XP_047296965.1
          XM_047441010.1 XP_047296966.1
          XM_047441011.1 XP_047296967.1
          XM_047441012.1 XP_047296968.1
          XM_047441013.1 XP_047296969.1
          XM_047441014.1 XP_047296970.1
          XM_047441015.1 XP_047296971.1
          AK226159.1
          AL581043.3
          BC136380.1 AAI36381.1
          BC136381.1 AAI36382.1
          BC144053.1 AAI44054.1
          D43968.1 BAA07903.1
          D43969.1 BAA07904.1
          D89788.1 BAA14020.1
          D89789.1 BAA14021.1
          D89790.1 BAA14022.1
          L34598.1 AAA51720.1
          X79549.1 CAA56092.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv5550749 Chr.21:34567808 - 34891808 on Build GRCh38 Duplication RUNX1-AS1 LINC01426 LINC00160 CLIC6 RCAN1 RUNX1
          nsv5539790 Chr.21:5227438 - 39583816 on Build GRCh38 Insertion KRTAP21-3 MIR10396A MIR6724-2 LINC00160 KRTAP8-1 CLDN17 LOC102723360 KRTAP13-3 KRTAP11-1 LOC107983987 GET1 USP16 MIS18A APP IL10RB KRTAP21-1 LINC00320 TIAM1-AS1 OLIG2 LOC102724354 GRIK1-AS1 MEMO1P1 KRTAP6-1 SMIM11 MIR4327 MIR155HG RPL23P2 LINC01423 C21orf140 LOC105379504 KRTAP25-1 ETS2 CYYR1 ERG ITSN1 MRAP EPCIP LOC105369306 LOC107987288 RNA28SN1 BRWD1-AS2 IFNAR2 LINC01425 KRTAP19-2 TPTE LINC00189 LINC02573 LINC00158 LOC102724428 SETD4-AS1 GRIK1 C21orf91 MIR548XHG BRWD1 CBR1-AS1 KRTAP19-5 LINC01673 LOC105377134 IFNAR1 BTG3 RNA45SN3 KRTAP6-3 DYRK1A SCAF4 IL10RB-DT CBR3 LINC00649 LINC01667 KCNE1 SOD1 RNA45SN1 MIR8069 LINC00314 MIR8069-1 TMPRSS15 HEMK2 LOC110091777 SNORA80A MIR99AHG LOC102723553 CLIC6 APP-DT LINC01666 RNA18SN1 ADAMTS1 LINC01700 LINC01436 LOC102724843 ATP5PF HMGN1 ANKRD20A11P SNORD74B KRTAP27-1 USP25 MIR125B2 KCNJ6-AS1 CBR1 MIR6508 CLDN14-AS1 SMIM34 IGHV1OR21-1 RNA45SN2 DSCR8 LINC03104 IFNAR2-IL10RB CBR3-AS1 LINC03105 LINC00515 NCAM2 ADAMTS5 ABCC13 NRIP1 MIR3118-1 OLIG1 LINC01687 LINC00317 SAMSN1 LINC01697 LINC01695 SON KRTAP7-1 SAMSN1-AS1 GRIK1-AS2 MIR6724-1 EVA1C LINC02920 KCNJ15 DSCR10 LINC00114 URB1 ASMER1 RBM11 LOC110091776 MRAP-AS1 MORC3 LINC01426 LINC00159 LOC101927345 KRTAP23-1 DOP1B RCAN1 LINC00307 BACH1 MIR99A DONSON LOC101928269 D21S2088E RNA5-8SN2 LINC01692 KRTAP13-1 MRPS6 CHODL LOC101930100 POTED CYYR1-AS1 TEKT4P2 LOC105372791 RUNX1-AS1 LINC00945 MIR6130 RNA28SN2 MRPL39 DSCR4 LOC106780825 LOC105379514 KRTAP20-1 RNA18SN3 ANKRD30BP2 RWDD2B LOC105379499 EPCIP-AS1 LOC101927745 LOC102723475 CH507-42P11.6 LOC102723451 LOC124905049 TTC3 LINC01548 KCNE2 JAM2 DSCR9 PAXBP1-AS1 KRTAP19-7 MIR155 SIM2 RUNX1 SETD4 GET1-SH3BGR MAP3K7CL LOC102724652 KRTAP6-2 KCNJ6 LOC100506403 RUNX1-IT1 RNA18SN2 LCA5L LOC101928107 CRYZL1 BACH1-IT2 TCP10L TTC3-AS1 LINC03147 KRTAP19-8 ETS2-AS1 MIR548X MIS18A-AS1 LOC102724594 NCOR1P4 RNA5-8SN1 KRTAP13-2 RNA5-8SN3 LOC105372753 LINC01689 LOC102724334 KRTAP20-2 KRTAP22-2 LOC100288590 RIPPLY3 KRTAP22-1 LOC105369308 MIR802 SH3BGR CXADR KRTAP24-1 LINC00308 LINC01690 LINC01549 BAGE2 LOC101928398 PSMG1 LOC105372793 CLDN14 MIR6501 CFAP298-TCP10L KRTAP15-1 KRTAP20-3 SOD1-DT SLC5A3 RNA28SN3 MIR3156-3 VPS26C MIRLET7C PIGP GABPA DNAJC28 LOC124905050 KRTAP19-6 MIR6724-4 TMEM50B KRTAP20-4 HLCS BRWD1-AS1 PPP1R2P2 CHAF1B LINC01674 HLCS-AS1 KRTAP19-1 GART LOC102724560 CLDN8 KRTAP19-4 LINC02943 LOC105372762 MIR4759 KRTAP19-3 LINC00113 LINC00161 C21orf91-OT1 PAXBP1 TIAM1 KRTAP13-4 CFAP298 MIR3648-1 HUNK LINC01683 BTG3-AS1 BACH1-IT3 CCT8 LINC00310 MIR6724-3 LOC101928126 LINC01684 LTN1 SYNJ1 ATP5PO GRIK1-AS3 MIR3648-2 MIR10396B KRTAP21-2 URB1-AS1 CYP4F29P KRTAP26-1 IFNGR2 LIPI HSPA13

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ossification
          chondrocyte differentiation
          myeloid leukocyte differentiation
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          regulation of plasminogen activation
          hemopoiesis
          myeloid cell differentiation
          neuron differentiation
          negative regulation of granulocyte differentiation
          positive regulation of granulocyte differentiation
          positive regulation of interleukin-2 production
          positive regulation of collagen biosynthetic process
          negative regulation of CD4-positive, alpha-beta T cell differentiation
          positive regulation of CD8-positive, alpha-beta T cell differentiation
          regulation of cell differentiation
          positive regulation of angiogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          peripheral nervous system neuron development
          positive regulation of developmental process
          positive regulation of multicellular organismal process
          regulation of cardiac muscle cell proliferation
          cardiac muscle tissue regeneration
          hematopoietic stem cell proliferation
          positive regulation of extracellular matrix organization
          regulation of connective tissue replacement
          regulation of multicellular organismal development
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcription corepressor binding
          transcription coactivator binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          calcium ion binding
          protein binding
          ATP binding
          protein homodimerization activity
          protein heterodimerization activity
          DNA-binding transcription factor binding
          sequence-specific double-stranded DNA binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline