Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05562880_cn
          See other SMARCB1 CNV Assays ›
          Gene Symbol
          SMARCB1
          Assay Reference Genome
          Location

          Chr.22:23787687 on build GRCh38
          Cytoband
          22q11.23
          Assay ID Hs05562880_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          6598

          Gene Name:

          SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1

          Gene Aliases:

          BAF47, CSS3, INI1, MRD15, PPP1R144, RDT, RTPS1, SNF5, SNF5L1, SWNTS1, Sfh1p, Snr1, hSNFS

          Location:

          Chr.22:23786931-23834518 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SMARCB1 NM_001007468.2 NP_001007469.1
          NM_001317946.1 NP_001304875.1
          NM_003073.4 NP_003064.2
          XM_011530345.2 XP_011528647.1
          AB017523.1 BAC77068.1
          AJ011737.1 CAA09758.1
          AJ011738.1 CAA09759.1
          AK021419.1
          AK024025.1 BAB14784.1
          AK299411.1
          AK303620.1
          AL389878.1
          AU143041.1
          BC117114.1
          BC143667.1
          BG705851.1
          CR456581.1 CAG30467.1
          HY013730.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv8039n54 Chr.22:23323169 - 24601506 on Build GRCh38 Gain PCAT14 ZDHHC8P1 MIF ADORA2A-AS1 GUCD1 DDTL SPECC1L CES5AP1 SMARCB1 SNRPD3 LRRC75B GSTTP1 DERL3 LOC388882 POM121L9P DDT C22orf15 ADORA2A LOC100652871 RGL4 ZNF70 VPREB3 GGT1 MIF-AS1 LOC101929374 GUSBP11 SLC2A11 DRICH1 UPB1 SUSD2 GSTT2 GGT5 MMP11 LOC107985559 LOC107985555 CABIN1 GSTT2B CHCHD10 SPECC1L-ADORA2A IGLL1
          esv3893461 Chr.22:23509514 - 23995877 on Build GRCh38 Gain PCAT14 LOC100652871 RGL4 ZNF70 VPREB3 MIF-AS1 MIF GUSBP11 SLC2A11 DRICH1 DDTL GSTT2 MMP11 SMARCB1 LOC107985559 GSTT2B DERL3 CHCHD10 DDT C22orf15 IGLL1
          nsv526567 Chr.22:23348138 - 24600663 on Build GRCh38 Gain PCAT14 ZDHHC8P1 MIF ADORA2A-AS1 GUCD1 DDTL SPECC1L CES5AP1 SMARCB1 SNRPD3 LRRC75B GSTTP1 DERL3 LOC388882 POM121L9P DDT C22orf15 ADORA2A LOC100652871 RGL4 ZNF70 VPREB3 GGT1 MIF-AS1 LOC101929374 GUSBP11 SLC2A11 DRICH1 UPB1 SUSD2 GSTT2 GGT5 MMP11 LOC107985559 LOC107985555 CABIN1 GSTT2B CHCHD10 SPECC1L-ADORA2A IGLL1
          esv2724041 Chr.22:23315914 - 24669376 on Build GRCh38 Deletion PCAT14 ZDHHC8P1 MIF ADORA2A-AS1 POM121L10P GUCD1 DDTL SPECC1L CES5AP1 SMARCB1 SNRPD3 LRRC75B GSTTP1 DERL3 LOC388882 BCRP3 POM121L9P DDT C22orf15 ADORA2A LOC100652871 RGL4 ZNF70 VPREB3 GGT1 MIF-AS1 LOC101929374 GUSBP11 SLC2A11 DRICH1 UPB1 SUSD2 GSTT2 GGT5 MMP11 LOC107985559 LOC107985555 CABIN1 GSTT2B BCR CHCHD10 SPECC1L-ADORA2A IGLL1
          nsv1062411 Chr.22:23407914 - 23922609 on Build GRCh38 Gain PCAT14 RGL4 ZNF70 VPREB3 MIF-AS1 MIF LOC101929374 GUSBP11 SLC2A11 DRICH1 MMP11 SMARCB1 DERL3 CHCHD10 LOC388882 C22orf15 IGLL1
          dgv4528n100 Chr.22:23322244 - 24682765 on Build GRCh38 Gain PCAT14 ZDHHC8P1 MIF ADORA2A-AS1 POM121L10P GUCD1 DDTL SPECC1L CES5AP1 SMARCB1 SNRPD3 LRRC75B GSTTP1 DERL3 LOC388882 BCRP3 POM121L9P DDT C22orf15 ADORA2A LOC100652871 RGL4 ZNF70 VPREB3 GGT1 MIF-AS1 LOC101929374 GUSBP11 SLC2A11 DRICH1 UPB1 SUSD2 GSTT2 GGT5 MMP11 LOC107985559 LOC107985555 CABIN1 GSTT2B CHCHD10 SPECC1L-ADORA2A IGLL1
          esv3893457 Chr.22:22657560 - 23995877 on Build GRCh38 Gain PCAT14 ZDHHC8P1 MIF DDTL IGLV4-3 MIR5571 CES5AP1 SMARCB1 IGLV3-19 IGLV3-21 IGLJ3 DERL3 LOC388882 DDT C22orf15 IGLL5 LOC100652871 RGL4 ZNF70 VPREB3 MIF-AS1 LOC101929374 GUSBP11 SLC2A11 RSPH14 MIR650 DRICH1 IGLC1 GSTT2 GNAZ MMP11 LOC107985559 LOC107985555 GSTT2B BCR CHCHD10 RAB36 IGLV2-5 IGLV3-25 FBXW4P1 IGLL1
          nsv1066891 Chr.22:22611045 - 24001082 on Build GRCh38 Gain PCAT14 ZDHHC8P1 MIF DDTL IGLV4-3 MIR5571 CES5AP1 SMARCB1 IGLV3-19 IGLV3-21 IGLJ3 GSTTP1 DERL3 LOC388882 DDT C22orf15 IGLL5 LOC100652871 RGL4 POM121L1P ZNF70 VPREB3 MIF-AS1 LOC101929374 GUSBP11 SLC2A11 RSPH14 MIR650 DRICH1 GGTLC2 IGLC1 GSTT2 GNAZ MMP11 LOC107985559 LOC107985555 GSTT2B BCR CHCHD10 RAB36 IGLV2-5 IGLV3-25 FBXW4P1 IGLL1
          esv3575443 Chr.22:23348201 - 24670505 on Build GRCh38 Gain PCAT14 ZDHHC8P1 MIF ADORA2A-AS1 POM121L10P GUCD1 DDTL SPECC1L CES5AP1 SMARCB1 SNRPD3 LRRC75B GSTTP1 DERL3 LOC388882 BCRP3 POM121L9P DDT C22orf15 ADORA2A LOC100652871 RGL4 ZNF70 VPREB3 GGT1 MIF-AS1 LOC101929374 GUSBP11 SLC2A11 DRICH1 UPB1 SUSD2 GSTT2 GGT5 MMP11 LOC107985559 LOC107985555 CABIN1 GSTT2B CHCHD10 SPECC1L-ADORA2A IGLL1

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          blastocyst hatching
          DNA repair
          nucleosome disassembly
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          cell cycle
          nervous system development
          negative regulation of cell proliferation
          DNA integration
          covalent chromatin modification
          cell differentiation
          single stranded viral RNA replication via double stranded DNA intermediate
          ATP-dependent chromatin remodeling
          positive regulation by host of viral transcription
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of histone H4 acetylation
          negative regulation of histone H3-K9 dimethylation
          negative regulation of histone H3-K9 trimethylation
          positive regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter
          positive regulation of glucose mediated signaling pathway
          positive regulation of histone H3-K9 acetylation
          regulation of histone H4-K16 acetylation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          RNA polymerase I CORE element sequence-specific DNA binding
          p53 binding
          DNA binding
          transcription coactivator activity
          protein binding
          Tat protein binding
          nucleosomal DNA binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0