Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05572706_cn
          See other SMARCB1 CNV Assays ›
          Gene Symbol
          SMARCB1
          Assay Reference Genome
          Location

          Chr.22:23788753 on build GRCh38
          Cytoband
          22q11.23
          Assay ID Hs05572706_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          6598

          Gene Name:

          SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1

          Gene Aliases:

          BAF47, CSS3, INI1, MRD15, PPP1R144, RDT, RTPS1, SNF5, SNF5L1, SWNTS1, Sfh1p, Snr1, hSNFS

          Location:

          Chr.22:23786931-23834518 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SMARCB1 NM_001007468.2 NP_001007469.1
          NM_001317946.1 NP_001304875.1
          NM_003073.4 NP_003064.2
          XM_011530345.2 XP_011528647.1
          AB017523.1 BAC77068.1
          AJ011737.1 CAA09758.1
          AJ011738.1 CAA09759.1
          AK021419.1
          AK024025.1 BAB14784.1
          AK299411.1
          AK303620.1
          AL389878.1
          AU143041.1
          BC117114.1
          BC143667.1
          BG705851.1
          CR456581.1 CAG30467.1
          HY013730.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv8039n54 Chr.22:23323169 - 24601506 on Build GRCh38 Gain CABIN1 DDT LOC100652871 DDTL DRICH1 ZDHHC8P1 GUSBP11 SPECC1L-ADORA2A LOC101929374 LRRC75B PCAT14 GSTTP1 GGT1 GGT5 SMARCB1 MIF GUCD1 SNRPD3 GSTT2 LOC388882 ADORA2A MMP11 CES5AP1 LOC107985555 UPB1 ADORA2A-AS1 GSTT2B CHCHD10 POM121L9P C22orf15 LOC107985559 RGL4 MIF-AS1 SUSD2 ZNF70 SPECC1L SLC2A11 IGLL1 VPREB3 DERL3
          esv3893461 Chr.22:23509514 - 23995877 on Build GRCh38 Gain DDT LOC100652871 DDTL GSTT2B CHCHD10 DRICH1 C22orf15 GUSBP11 LOC107985559 RGL4 PCAT14 MIF-AS1 SMARCB1 MIF GSTT2 ZNF70 MMP11 SLC2A11 IGLL1 VPREB3 DERL3
          nsv526567 Chr.22:23348138 - 24600663 on Build GRCh38 Gain CABIN1 DDT LOC100652871 DDTL DRICH1 ZDHHC8P1 GUSBP11 SPECC1L-ADORA2A LOC101929374 LRRC75B PCAT14 GSTTP1 GGT1 GGT5 SMARCB1 MIF GUCD1 SNRPD3 GSTT2 LOC388882 ADORA2A MMP11 CES5AP1 LOC107985555 UPB1 ADORA2A-AS1 GSTT2B CHCHD10 POM121L9P C22orf15 LOC107985559 RGL4 MIF-AS1 SUSD2 ZNF70 SPECC1L SLC2A11 IGLL1 VPREB3 DERL3
          esv2724041 Chr.22:23315914 - 24669376 on Build GRCh38 Deletion CABIN1 DDT LOC100652871 DDTL DRICH1 ZDHHC8P1 GUSBP11 SPECC1L-ADORA2A LOC101929374 LRRC75B PCAT14 GSTTP1 GGT1 GGT5 SMARCB1 MIF GUCD1 SNRPD3 GSTT2 LOC388882 ADORA2A MMP11 CES5AP1 BCRP3 POM121L10P LOC107985555 UPB1 ADORA2A-AS1 GSTT2B CHCHD10 POM121L9P C22orf15 LOC107985559 RGL4 MIF-AS1 SUSD2 ZNF70 BCR SPECC1L SLC2A11 IGLL1 VPREB3 DERL3
          nsv1062411 Chr.22:23407914 - 23922609 on Build GRCh38 Gain CHCHD10 DRICH1 C22orf15 GUSBP11 LOC101929374 RGL4 PCAT14 MIF-AS1 SMARCB1 MIF LOC388882 ZNF70 MMP11 SLC2A11 IGLL1 VPREB3 DERL3
          dgv4528n100 Chr.22:23322244 - 24682765 on Build GRCh38 Gain CABIN1 DDT LOC100652871 DDTL DRICH1 ZDHHC8P1 GUSBP11 SPECC1L-ADORA2A LOC101929374 LRRC75B PCAT14 GSTTP1 GGT1 GGT5 SMARCB1 MIF GUCD1 SNRPD3 GSTT2 LOC388882 ADORA2A MMP11 CES5AP1 BCRP3 POM121L10P LOC107985555 UPB1 ADORA2A-AS1 GSTT2B CHCHD10 POM121L9P C22orf15 LOC107985559 RGL4 MIF-AS1 SUSD2 ZNF70 SPECC1L SLC2A11 IGLL1 VPREB3 DERL3
          esv3893457 Chr.22:22657560 - 23995877 on Build GRCh38 Gain IGLC1 DDT LOC100652871 DDTL DRICH1 ZDHHC8P1 GUSBP11 LOC101929374 FBXW4P1 PCAT14 IGLV3-19 SMARCB1 MIF GSTT2 LOC388882 MMP11 MIR5571 CES5AP1 MIR650 IGLV3-21 IGLJ3 LOC107985555 IGLV2-5 GSTT2B CHCHD10 C22orf15 IGLV3-25 RAB36 LOC107985559 RGL4 MIF-AS1 GNAZ IGLV4-3 RSPH14 ZNF70 BCR SLC2A11 IGLL1 IGLL5 VPREB3 DERL3
          nsv1066891 Chr.22:22611045 - 24001082 on Build GRCh38 Gain IGLC1 DDT LOC100652871 DDTL DRICH1 ZDHHC8P1 GGTLC2 GUSBP11 LOC101929374 FBXW4P1 PCAT14 GSTTP1 IGLV3-19 SMARCB1 MIF GSTT2 LOC388882 MMP11 POM121L1P MIR5571 CES5AP1 MIR650 IGLV3-21 IGLJ3 LOC107985555 IGLV2-5 GSTT2B CHCHD10 C22orf15 IGLV3-25 RAB36 LOC107985559 RGL4 MIF-AS1 GNAZ IGLV4-3 RSPH14 ZNF70 BCR SLC2A11 IGLL1 IGLL5 VPREB3 DERL3
          esv3575443 Chr.22:23348201 - 24670505 on Build GRCh38 Gain CABIN1 DDT LOC100652871 DDTL DRICH1 ZDHHC8P1 GUSBP11 SPECC1L-ADORA2A LOC101929374 LRRC75B PCAT14 GSTTP1 GGT1 GGT5 SMARCB1 MIF GUCD1 SNRPD3 GSTT2 LOC388882 ADORA2A MMP11 CES5AP1 BCRP3 POM121L10P LOC107985555 UPB1 ADORA2A-AS1 GSTT2B CHCHD10 POM121L9P C22orf15 LOC107985559 RGL4 MIF-AS1 SUSD2 ZNF70 SPECC1L SLC2A11 IGLL1 VPREB3 DERL3

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          blastocyst hatching
          DNA repair
          nucleosome disassembly
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          cell cycle
          nervous system development
          negative regulation of cell proliferation
          DNA integration
          covalent chromatin modification
          cell differentiation
          single stranded viral RNA replication via double stranded DNA intermediate
          ATP-dependent chromatin remodeling
          positive regulation by host of viral transcription
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of histone H4 acetylation
          negative regulation of histone H3-K9 dimethylation
          negative regulation of histone H3-K9 trimethylation
          positive regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter
          positive regulation of glucose mediated signaling pathway
          positive regulation of histone H3-K9 acetylation
          regulation of histone H4-K16 acetylation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          RNA polymerase I CORE element sequence-specific DNA binding
          p53 binding
          DNA binding
          transcription coactivator activity
          protein binding
          Tat protein binding
          nucleosomal DNA binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0