Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05851725_cn
          See other ILKAP CNV Assays ›
          Gene Symbol
          ILKAP
          Assay Reference Genome
          Location

          Chr.2:238175600 on build GRCh38
          Cytoband
          Assay ID Hs05851725_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          80895

          Gene Name:

          ILK associated serine/threonine phosphatase

          Gene Aliases:

          ILKAP2, ILKAP3, PP2C-DELTA, PP2CD, PPM1O

          Location:

          Chr.2:238170402-238203695 on Build GRCh38

          Assay Gene Location:

          Within Intron 9
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ILKAP NM_030768.3 NP_110395.1
          XM_006712784.2 XP_006712847.1
          XM_017005057.2 XP_016860546.1
          XM_017005058.2 XP_016860547.1
          AK001043.1 BAG50851.1
          AK055417.1
          AL136850.1 CAB66784.1
          AY024365.1 AAK07736.1
          BC006576.2 AAH06576.1
          CR533533.1 CAG38564.1
          FJ491380.1 ACL54845.1
          HQ658059.1 AEI30977.1
          HQ658060.1 AEI30978.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv4080148 Chr.2:238167624 - 238191446 on Build GRCh38 Duplication ILKAP ERFE
          nsv508894 Chr.2:238076839 - 238177028 on Build GRCh38 Insertion KLHL30 ILKAP ESPNL SCLY UBE2F-SCLY ERFE
          nsv1000252 Chr.2:235899999 - 238766417 on Build GRCh38 Loss KLHL30 RAB17 ILKAP HES6 SCLY IQCA1-AS1 MIR6811 AGAP1 PER2 COPS8 UBE2F-SCLY ASB18 DRC11 GBX2 GBX2-AS1 LOC93463 LRRFIP1 RAB17-DT TRAF3IP1 RAMP1 PRLH ESPNL COL6A3 ERFE MLPH TARDBPP3 UBE2F LINC02610 RBM44 COPS8-DT ACKR3 ASB1 LINC01107

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          phosphoprotein phosphatase activity
          protein serine/threonine phosphatase activity
          protein binding
          hydrolase activity
          cation binding
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline