Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05910046_cn
          See other EDNRA CNV Assays ›
          Gene Symbol
          EDNRA
          Assay Reference Genome
          Location

          Chr.4:147541267 on build GRCh38
          Cytoband
          Assay ID Hs05910046_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1909

          Gene Name:

          endothelin receptor type A

          Gene Aliases:

          ET-A, ETA, ETA-R, ETAR, ETRA, MFDA, hET-AR

          Location:

          Chr.4:147481097-147544954 on Build GRCh38

          Assay Gene Location:

          Within Intron 8
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          EDNRA NM_001166055.2 NP_001159527.1
          NM_001957.4 NP_001948.1
          NR_045958.2
          NR_148963.2
          NR_148964.2
          AK304451.1 BAG65268.1
          AK312812.1 BAG35670.1
          AK315931.1 BAH14302.1
          AY275462.1 AAP32294.1
          BC022511.1 AAH22511.1
          D90348.1 BAA14359.1
          L06622.1 AAA58447.1
          S45956.1 AAB23644.1
          S57498.1 AAB25530.2
          S63938.1 AAB20278.1
          S67127.1 AAB20407.1
          S81539.1 AAB36325.1
          S81542.1 AAB36326.1
          S81545.1 AAB36327.1
          X61950.1 CAA43953.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv830110 Chr.4:147510973 - 147673968 on Build GRCh38 Loss EDNRA LINC02507 TMEM184C-DT PRMT9 TMEM184C
          nsv6387866 Chr.4:147538037 - 147970462 on Build GRCh38 Duplication EDNRA MIR4799 ARHGAP10 LINC02507 TMEM184C-DT PRMT9 TMEM184C
          nsv830109 Chr.4:147373909 - 147547453 on Build GRCh38 Loss EDNRA

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          angiogenesis
          patterning of blood vessels
          response to hypoxia
          in utero embryonic development
          blood vessel remodeling
          response to amphetamine
          regulation of heart rate
          heart process
          glomerular filtration
          cardiac chamber formation
          left ventricular cardiac muscle tissue morphogenesis
          atrial cardiac muscle tissue development
          cardiac neural crest cell migration involved in outflow tract morphogenesis
          noradrenergic neuron differentiation
          cellular calcium ion homeostasis
          smooth muscle contraction
          mitochondrion organization
          signal transduction
          G-protein coupled receptor signaling pathway
          activation of adenylate cyclase activity
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          axon guidance
          heart development
          respiratory gaseous exchange
          regulation of blood pressure
          cell proliferation
          response to wounding
          gene expression
          regulation of glucose transport
          neural crest cell development
          neural crest cell differentiation
          neural crest cell fate commitment
          artery smooth muscle contraction
          vascular smooth muscle contraction
          neuron remodeling
          heparin metabolic process
          thyroid gland development
          neuron projection development
          glomerulus development
          cellular response to oxidative stress
          embryonic heart tube development
          aorta development
          vasoconstriction
          norepinephrine metabolic process
          middle ear morphogenesis
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          cellular response to human chorionic gonadotropin stimulus
          developmental pigmentation
          enteric nervous system development
          sympathetic nervous system development
          axon extension
          embryonic skeletal system development
          neuromuscular process
          sodium ion homeostasis
          canonical Wnt signaling pathway
          head development
          face development
          axonogenesis involved in innervation
          establishment of endothelial barrier
          pharyngeal arch artery morphogenesis
          renal sodium ion absorption
          calcium ion transmembrane transport
          cellular response to follicle-stimulating hormone stimulus
          cellular response to luteinizing hormone stimulus
          protein transmembrane transport
          glomerular endothelium development
          glomerular visceral epithelial cell differentiation
          endothelin receptor signaling pathway
          endothelin receptor signaling pathway involved in heart process
          renal albumin absorption
          vascular smooth muscle cell development
          mesenchymal cell apoptotic process
          cellular response to toxic substance
          sympathetic neuron axon guidance
          cAMP/PKA signal transduction
          semaphorin-plexin signaling pathway involved in axon guidance
          glomerular visceral epithelial cell apoptotic process
          meiotic cell cycle process involved in oocyte maturation
          cranial skeletal system development
          response to acetylcholine
          regulation of protein localization to cell leading edge
          phosphatidylinositol phospholipase C activity
          G-protein coupled receptor activity
          endothelin receptor activity
          protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline