Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06157041_cn
          See other CASP8AP2 CNV Assays ›
          Gene Symbol
          CASP8AP2
          Assay Reference Genome
          Location

          Chr.6:89841040 on build GRCh38
          Cytoband
          Assay ID Hs06157041_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          9994

          Gene Name:

          caspase 8 associated protein 2

          Gene Aliases:

          CED-4, FLASH, RIP25

          Location:

          Chr.6:13399-72124 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CASP8AP2 NM_001137667.2 NP_001131139.1
          NM_001137668.2 NP_001131140.1
          NM_012115.4 NP_036247.1
          AF154415.1 AAF03367.1
          BC056685.1 AAH56685.1
          BC132830.1 AAI32831.1
          BP192859.1
          BP363682.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv5391 Chr.6:89837037 - 89869200 on Build GRCh38 Insertion CASP8AP2
          nsv604125 Chr.6:89751706 - 90189842 on Build GRCh38 Gain BACH2 GJA10 CASP8AP2 MDN1
          esv3890901 Chr.6:89204468 - 90033564 on Build GRCh38 Gain ANKRD6 LYRM2 BACH2 GJA10 GABRR2 UBE2J1 CASP8AP2 GABRR1 MDN1 MDN1-AS1 RRAGD
          nsv6405432 Chr.6:89376654 - 90197092 on Build GRCh38 Duplication ANKRD6 LYRM2 BACH2 GJA10 CASP8AP2 MDN1 MDN1-AS1 RRAGD

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          signal transduction
          extrinsic apoptotic signaling pathway via death domain receptors
          Fas signaling pathway
          negative regulation of transcription, DNA-templated
          cellular response to mechanical stimulus
          apoptotic signaling pathway
          transcription corepressor activity
          death receptor binding
          protein binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          peptidase activator activity involved in apoptotic process
          SUMO polymer binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline