Search
Search

Entrez Gene ID: | 28999 |
Gene Name: | Kruppel like factor 15 |
Gene Aliases: |
KKLF |
Location: |
Chr.3:126288121-126357393 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF15 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591500 | Chr.3:126296824 - 126357010 on Build GRCh38 | Loss |
|