Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06911453_cn
          See other MTAP CNV Assays ›
          Gene Symbol
          MTAP
          Assay Reference Genome
          Location

          Chr.9:21807324 on build GRCh38
          Cytoband
          Assay ID Hs06911453_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          4507

          Gene Name:

          methylthioadenosine phosphorylase

          Gene Aliases:

          BDMF, DMSFH, DMSMFH, HEL-249, LGMBF, MSAP, c86fus

          Location:

          Chr.9:21802636-21941115 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MTAP NM_001396040.1 NP_001382969.1
          NM_001396041.1 NP_001382970.1
          NM_001396042.1 NP_001382971.1
          NM_001396043.1 NP_001382972.1
          NM_001396044.1 NP_001382973.1
          NM_001396045.1 NP_001382974.1
          NM_002451.4 NP_002442.2
          NR_173242.1
          AF109294.1 AAD19641.1
          AK300592.1 BAG62290.1
          AK309365.1
          BC012316.1 AAH12316.1
          BC026106.1 AAH26106.1
          CR541670.1 CAG46471.1
          CR541710.1 CAG46511.1
          FJ224338.1 ACI46030.1
          GQ891397.1 ADO22259.1
          HE654772.1 CCF77345.1
          HE654773.1 CCF77346.1
          HE654774.1 CCF77347.1
          HE654775.1 CCF77348.1
          HE654776.1 CCF77349.1
          HE654777.1 CCF77350.1
          L40432.1 AAG38871.1
          U22233.1 AAA81646.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv4184515 Chr.9:21703459 - 22598303 on Build GRCh38 Duplication CDKN2A-AS1 CDKN2B-AS1 DMRTA1 CDKN2B CDKN2A MTAP
          nsv6000594 Chr.9:21677724 - 23021905 on Build GRCh38 Duplication CDKN2A-AS1 CDKN2B-AS1 DMRTA1 CDKN2B CDKN2A MTAP LINC01239
          nsv1026981 Chr.9:21748203 - 21811698 on Build GRCh38 Gain MTAP
          esv2764150 Chr.9:20659506 - 22349184 on Build GRCh38 Gain+Loss CDKN2A-AS1 IFNA13 IFNA10 MIR31HG MIR491 IFNA17 IFNA16 IFNB1 CDKN2B-AS1 IFNA14 IFNA8 CDKN2A IFNA21 HACD4 IFNA7 IFNW1 CDKN2B IFNA22P IFNA1 KLHL9 SNORA30B IFNA4 IFNA2 IFNA5 IFNA6 IFNE MTAP FOCAD-AS1 FOCAD MIR31
          nsv8426 Chr.9:21763811 - 22176201 on Build GRCh38 Loss CDKN2A-AS1 CDKN2B-AS1 CDKN2B CDKN2A MTAP
          esv3619972 Chr.9:21806006 - 21817430 on Build GRCh38 Loss MTAP
          nsv5479271 Chr.9:21806005 - 21817449 on Build GRCh38 Deletion MTAP

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs75002873] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          purine ribonucleoside salvage
          nicotinamide riboside catabolic process
          nucleoside metabolic process
          L-methionine biosynthetic process from methylthioadenosine
          methylation
          response to testosterone
          catalytic activity
          phosphorylase activity
          protein binding
          transferase activity
          transferase activity, transferring glycosyl groups
          transferase activity, transferring pentosyl groups
          S-methyl-5-thioadenosine phosphorylase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline