Search
Search

Entrez Gene ID: | 113675 |
Gene Name: | serine dehydratase like |
Gene Aliases: |
SDH 2, SDHL, SDS-RS1, TDH, cSDH |
Location: |
Chr.12:113422380-113438276 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDSL | NM_001304993.2 | NP_001291922.1 | ||
| NM_138432.4 | NP_612441.1 | |||
| XM_005253831.6 | XP_005253888.1 | |||
| XM_017018763.2 | 1 | 322 | XP_016874252.1 | |
| AF134473.1 | AAP97250.1 | |||
| BC009849.2 | AAH09849.1 | |||
| BC091479.1 | AAH91479.1 |
DGV Version: |
Release date: 2025-12-01, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss |
|
| esv3892265 | Chr.12:113099321 - 113527932 on Build GRCh38 | Gain |
|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|
Set Membership: |
|