Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs06976599_cn
See other PARPBP CNV Assays ›
Gene Symbol
PARPBP
Assay Reference Genome
Location
Chr.12:102126016 on build GRCh38
Cytoband
12q23.2
Assay ID
Hs06976599_cn
Size
S: 360 rxns
M: 750 rxns
L: 2,900 rxns
Availability
Made To Order
Catalog #
4400291
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Target Gene Details
Entrez Gene ID:
55010
Gene Name:
PARP1 binding protein
Gene Aliases:
AROM, C12orf48, PARI
Location:
Chr.12:102120171-102197520 on Build GRCh38
Assay Gene Location:
Within Intron 2
Gene Symbol
Transcript Accession
Exon Location
Assay Transcript Location
Protein ID
PARPBP
NM_001319988.1
NP_001306917.1
NM_001319993.1
NP_001306922.1
NM_001319994.1
NP_001306923.1
NM_001319995.1
NP_001306924.1
NM_001319996.1
NP_001306925.1
NM_017915.4
NP_060385.3
NR_135123.1
XM_011538509.1
XP_011536811.1
XM_011538514.2
XP_011536816.1
XM_017019534.1
XP_016875023.1
XM_017019536.1
XP_016875025.1
XM_017019537.1
XP_016875026.1
XM_017019538.1
XP_016875027.1
XM_017019539.1
XP_016875028.1
XM_017019540.1
XP_016875029.1
XM_017019541.1
XP_016875030.1
XM_017019542.1
XP_016875031.1
XM_017019543.1
XP_016875032.1
XM_017019544.1
XP_016875033.1
AF274940.1
AAK07517.1
AK000648.1
BAA91306.1
AK300041.1
AK302724.1
AK303571.1
BC018903.2
AAH18903.1
BC030962.1
BC050696.1
AAH50696.1
BC070270.1
AAH70270.1
BC098313.1
AAH98313.1
BC099734.1
BC099844.1
BG285036.1
DB260405.1
Back To Top
More Information
Set Membership:
Intragenic
Intronic
Non-exonic
non-DGV Variation
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
negative regulation of double-strand break repair via homologous recombination
DNA binding
protein binding
Back To Top
Related Products
TaqMan
®
Genotyping Master Mix
TaqPath
®
ProAmp
®
Master Mix
TaqMan
®
Universal Master Mix II, no UNG
TaqMan
®
Gene Expression Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Universal PCR Master Mix, No AmpErase UNG
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline