Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07179640_cn
          See other MACROD2 CNV Assays ›
          Gene Symbol
          MACROD2
          Assay Reference Genome
          Location

          Chr.20:14583161 on build GRCh38
          Cytoband
          Assay ID Hs07179640_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          140733

          Gene Name:

          mono-ADP ribosylhydrolase 2

          Gene Aliases:

          C20orf133, C2orf133

          Location:

          Chr.20:13995516-16053197 on Build GRCh38

          Assay Gene Location:

          Within Intron 4
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MACROD2 NM_001351661.2 NP_001338590.1
          NM_001351663.2 NP_001338592.1
          NM_080676.6 NP_542407.2
          AK131348.1 BAD18504.1

          Target Gene Details

          Entrez Gene ID:

          140848

          Gene Name:

          MACROD2 intronic transcript 1

          Gene Aliases:

          C20orf68, NCRNA00227, bA467D7.4

          Location:

          Chr.20:14554384-14628908 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          MACROD2-IT1 NR_104193.2
          CR747489.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          dgv7464n54 Chr.20:14549011 - 14607522 on Build GRCh38 Loss MACROD2-IT1 MACROD2
          nsv4287029 Chr.20:14581027 - 14585815 on Build GRCh38 Duplication MACROD2-IT1 MACROD2
          nsv3113317 Chr.20:14512089 - 14583844 on Build GRCh38 Deletion MACROD2-IT1 MACROD2
          dgv1422n166 Chr.20:14567859 - 14822341 on Build GRCh38 Deletion MACROD2-IT1 MACROD2
          nsv1064721 Chr.20:14404285 - 14667577 on Build GRCh38 Loss MACROD2-IT1 MACROD2
          nsv1061504 Chr.20:14537718 - 14585075 on Build GRCh38 Loss MACROD2-IT1 MACROD2
          nsv4284681 Chr.20:14541458 - 14600276 on Build GRCh38 Duplication MACROD2-IT1 MACROD2
          nsv3219574 Chr.20:14566112 - 14626057 on Build GRCh38 Deletion MACROD2-IT1 MACROD2
          nsv833926 Chr.20:14429507 - 14591690 on Build GRCh38 Gain MACROD2-IT1 MACROD2
          nsv6527658 Chr.20:14532149 - 14615660 on Build GRCh38 Deletion MACROD2-IT1 MACROD2
          dgv241n21 Chr.20:14573099 - 14593321 on Build GRCh38 Loss MACROD2-IT1 MACROD2
          nsv4287587 Chr.20:14511361 - 14584427 on Build GRCh38 Deletion MACROD2-IT1 MACROD2
          nsv4272536 Chr.20:14518421 - 14625243 on Build GRCh38 Deletion MACROD2-IT1 MACROD2
          dgv4380n223 Chr.20:14576230 - 14677212 on Build GRCh38 Deletion MACROD2-IT1 MACROD2
          nsv517905 Chr.20:14556162 - 14646199 on Build GRCh38 Loss MACROD2-IT1 MACROD2
          nsv585450 Chr.20:14568030 - 14629643 on Build GRCh38 Loss MACROD2-IT1 MACROD2
          dgv4250n100 Chr.20:14543758 - 14705651 on Build GRCh38 Loss MACROD2-IT1 MACROD2
          nsv1055269 Chr.20:14580246 - 14665555 on Build GRCh38 Loss MACROD2-IT1 MACROD2
          esv2763220 Chr.20:14580258 - 14615047 on Build GRCh38 Gain MACROD2-IT1 MACROD2
          dgv1423n166 Chr.20:14574066 - 14695632 on Build GRCh38 Deletion MACROD2-IT1 MACROD2
          nsv6523732 Chr.20:14454766 - 14631942 on Build GRCh38 Deletion MACROD2-IT1 MACROD2
          nsv4276381 Chr.20:14553045 - 14654705 on Build GRCh38 Deletion MACROD2-IT1 MACROD2
          nsv6531720 Chr.20:14561245 - 14705822 on Build GRCh38 Deletion MACROD2-IT1 MACROD2
          nsv6048197 Chr.20:14569609 - 14613350 on Build GRCh38 Deletion MACROD2-IT1 MACROD2

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular response to DNA damage stimulus
          brain development
          response to bacterium
          purine nucleoside metabolic process
          protein de-ADP-ribosylation
          peptidyl-glutamate ADP-deribosylation
          hydrolase activity
          hydrolase activity, acting on glycosyl bonds
          deacetylase activity
          O-acetyl-ADP-ribose deacetylase activity
          ADP-ribosylglutamate hydrolase activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline