Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07401381_cn
          See other ATP6V0C CNV Assays ›
          Gene Symbol
          ATP6V0C
          Assay Reference Genome
          Location

          Chr.16:2516709 on build GRCh38
          Cytoband
          16p13.3
          Assay ID Hs07401381_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          527

          Gene Name:

          ATPase H+ transporting V0 subunit c

          Gene Aliases:

          ATP6C, ATP6L, ATPL, VATL, VPPC, Vma3

          Location:

          Chr.16:2513726-2520223 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ATP6V0C NM_001198569.1 NP_001185498.1
          NM_001694.3 NP_001685.1
          AW161158.1
          BC004537.2 AAH04537.1
          BC007389.2 AAH07389.1
          BC007759.2 AAH07759.1
          BC009290.2 AAH09290.1
          BI548787.1
          BT007155.1 AAP35819.1
          CR541930.1 CAG46728.1
          CR541951.1 CAG46749.1
          M62762.1 AAA60039.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2422427 Chr.16:10001 - 2723164 on Build GRCh38 Duplication C1QTNF8 MIR940 RNF151 PIGQ LOC105371047 SNORD60 SSTR5-AS1 MIR6511B1 METTL26 LMF1 MCRIP2 PGP TPSD1 LOC652276 TPSB2 HAGH TELO2 TRAF7 TMEM204 NOXO1 DECR2 LOC105371049 NPW RPL23AP5 SNORA64 SSTR5 CASKIN1 LOC101929280 TBL3 MPG WFIKKN1 GFER ZNF598 LOC106660606 LINC00235 CHTF18 RAB26 SNRNP25 METRN SNORA78 FAHD1 MAPK8IP3 DDX11L10 WDR24 CACNA1H HBM LOC105371184 HBQ1 CAPN15 NUBP2 SPSB3 TPSAB1 PKD1 ATP6V0C MSLN FBXL16 LOC105371046 RHOT2 TPSG1 C16orf59 CLCN7 LOC100134368 LOC107987233 NME3 MIR4717 MRPL28 LINC00254 MIR3177 RGS11 FAM234A HBZ HBA1 BAIAP3 CCDC154 AMDHD2 JMJD8 MEIOB POLR3K CRAMP1 RPUSD1 MIR662 MIR3178 SYNGR3 PDPK1 MIR3677 FLJ42627 ECI1 LMF1-AS1 MIR1225 PRR35 NDUFB10 NTHL1 TSR3 RHBDF1 MSRB1 UBE2I PRR25 NARFL IGFALS GNG13 MIR5587 RHBDL1 SLC9A3R2 MRPS34 CCDC78 RAB11FIP3 LOC107984876 MIR6767 TMEM8A DNASE1L2 IFT140 LOC105371045 STUB1 LOC729652 SNHG19 E4F1 NHLRC4 PRSS27 RPL3L RNPS1 TSC2 MIR3176 NTN3 UNKL LOC100287175 SNORA10 HS3ST6 C16orf91 RPS2 NME4 TBC1D24 SOX8 HAGHL ARHGDIG ABCA3 LOC105371038 SNHG9 WDR90 CEMP1 NPRL3 WASIR2 EME2 AXIN1 MIR3180-5 CCNF FAM173A PRSS29P PTX4 MIR6859-4 BRICD5 ERVK13-1 GNPTG HN1L MLST8 RAB40C MIR6768 LUC7L KCTD5 MIR4516 ABCA17P PDIA2 HBA2
          esv2760067 Chr.16:2486824 - 2820535 on Build GRCh38 Gain+Loss PDPK1 CEMP1 FLJ42627 PRSS27 PRSS21 PRSS33 ERVK13-1 ATP6V0C SRRM2-AS1 KCTD5 SRRM2 AMDHD2 TBC1D24 LOC652276 TCEB2 PRSS41 MIR3178
          nsv833122 Chr.16:2501717 - 2663937 on Build GRCh38 Loss PDPK1 CEMP1 FLJ42627 AMDHD2 TBC1D24 LOC652276 ERVK13-1 ATP6V0C MIR3178
          nsv1052260 Chr.16:2074546 - 2629655 on Build GRCh38 Gain PDPK1 E4F1 MIR3677 ECI1 MIR1225 RNPS1 MIR940 SNORD60 TSC2 PKD1 MIR6511B1 NTN3 ATP6V0C PGP TBC1D24 LOC652276 C16orf59 ABCA3 TRAF7 CEMP1 LOC105371049 MIR4717 MIR3180-5 CASKIN1 CCNF BRICD5 MLST8 MIR6768 LOC106660606 RAB26 MIR6767 AMDHD2 DNASE1L2 MIR4516 ABCA17P LOC729652 SNHG19 MIR3178
          nsv833121 Chr.16:2360975 - 2518228 on Build GRCh38 Loss MIR6768 MIR6767 TBC1D24 CCNF C16orf59 ABCA17P LOC729652 NTN3 ATP6V0C

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          ATP synthase

          Gene Ontology Categories:

          Function(s) Process(es)

          lysosomal lumen acidification
          insulin receptor signaling pathway
          ATP hydrolysis coupled proton transport
          proton transport
          viral process
          regulation of macroautophagy
          positive regulation of Wnt signaling pathway
          transferrin transport
          ion transmembrane transport
          phagosome acidification
          protein binding
          ubiquitin protein ligase binding
          proton-transporting ATP synthase activity, rotational mechanism
          proton-transporting ATPase activity, rotational mechanism

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline