Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07401393_cn
          See other ATP6V0C CNV Assays ›
          Gene Symbol
          ATP6V0C
          Assay Reference Genome
          Location

          Chr.16:2516917 on build GRCh38
          Cytoband
          16p13.3
          Assay ID Hs07401393_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          527

          Gene Name:

          ATPase H+ transporting V0 subunit c

          Gene Aliases:

          ATP6C, ATP6L, ATPL, VATL, VPPC, Vma3

          Location:

          Chr.16:2513726-2520223 on Build GRCh38

          Assay Gene Location:

          Within Intron 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ATP6V0C NM_001198569.1 NP_001185498.1
          NM_001694.3 NP_001685.1
          AW161158.1
          BC004537.2 AAH04537.1
          BC007389.2 AAH07389.1
          BC007759.2 AAH07759.1
          BC009290.2 AAH09290.1
          BI548787.1
          BT007155.1 AAP35819.1
          CR541930.1 CAG46728.1
          CR541951.1 CAG46749.1
          M62762.1 AAA60039.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2422427 Chr.16:10001 - 2723164 on Build GRCh38 Duplication MIR6767 MIR3180-5 WASIR2 BRICD5 MEIOB SOX8 CHTF18 METTL26 RPL23AP5 PDPK1 RPS2 MIR3677 SNORA78 CRAMP1 MIR5587 LOC107984876 PTX4 RHOT2 SNHG9 SPSB3 MIR6511B1 LOC729652 MRPS34 CCNF ABCA17P NTN3 ABCA3 PRSS29P KCTD5 LOC105371047 LOC105371184 TELO2 C1QTNF8 RHBDL1 MIR4717 RGS11 FAM234A NARFL NUBP2 MSRB1 FLJ42627 FAHD1 LOC105371046 CEMP1 TSC2 TMEM204 FAM173A MAPK8IP3 C16orf91 NPW LOC107987233 MIR3178 JMJD8 TBL3 RAB11FIP3 EME2 MIR940 GNG13 CASKIN1 C16orf59 LUC7L RNF151 MCRIP2 WDR24 LMF1 HN1L IFT140 DDX11L10 CCDC78 HBZ WFIKKN1 RHBDF1 MLST8 SNHG19 TPSD1 RAB40C CAPN15 LOC106660606 SSTR5 HBM DNASE1L2 NHLRC4 HBA1 ECI1 LOC105371045 TPSB2 PRR25 AMDHD2 NPRL3 GFER ZNF598 RPL3L MIR6768 CCDC154 HBA2 LOC105371038 SYNGR3 LMF1-AS1 TRAF7 RPUSD1 MSLN MIR3176 METRN E4F1 UBE2I HBQ1 CLCN7 TMEM8A ATP6V0C LINC00235 LOC100287175 TPSG1 NME3 MIR662 PRR35 WDR90 GNPTG LOC101929280 POLR3K SNORA10 MRPL28 HS3ST6 SNORA64 LINC00254 RNPS1 MPG TPSAB1 LOC652276 DECR2 AXIN1 MIR1225 NDUFB10 TBC1D24 MIR4516 FBXL16 NME4 IGFALS CACNA1H NOXO1 PIGQ TSR3 PRSS27 HAGH NTHL1 MIR3177 LOC100134368 SLC9A3R2 SNORD60 SNRNP25 SSTR5-AS1 HAGHL STUB1 UNKL LOC105371049 PGP BAIAP3 ARHGDIG ERVK13-1 RAB26 MIR6859-4 PKD1 PDIA2
          esv2760067 Chr.16:2486824 - 2820535 on Build GRCh38 Gain+Loss CEMP1 LOC652276 TCEB2 PDPK1 KCTD5 TBC1D24 SRRM2 AMDHD2 ERVK13-1 PRSS33 ATP6V0C MIR3178 PRSS21 PRSS27 PRSS41 SRRM2-AS1 FLJ42627
          nsv833122 Chr.16:2501717 - 2663937 on Build GRCh38 Loss TBC1D24 AMDHD2 ERVK13-1 ATP6V0C CEMP1 MIR3178 LOC652276 PDPK1 FLJ42627
          nsv1052260 Chr.16:2074546 - 2629655 on Build GRCh38 Gain MIR6767 SNHG19 MIR3180-5 BRICD5 RNPS1 CEMP1 TSC2 LOC106660606 DNASE1L2 LOC652276 PDPK1 ECI1 MIR1225 MIR3677 TBC1D24 AMDHD2 MIR4516 MIR3178 MIR6768 MIR940 MIR6511B1 TRAF7 CASKIN1 LOC729652 C16orf59 SNORD60 CCNF ABCA17P NTN3 ABCA3 LOC105371049 PGP E4F1 RAB26 ATP6V0C MIR4717 PKD1 MLST8
          nsv833121 Chr.16:2360975 - 2518228 on Build GRCh38 Loss TBC1D24 MIR6767 ATP6V0C LOC729652 C16orf59 CCNF ABCA17P NTN3 MIR6768

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          ATP synthase

          Gene Ontology Categories:

          Function(s) Process(es)

          lysosomal lumen acidification
          insulin receptor signaling pathway
          ATP hydrolysis coupled proton transport
          proton transport
          viral process
          regulation of macroautophagy
          positive regulation of Wnt signaling pathway
          transferrin transport
          ion transmembrane transport
          phagosome acidification
          protein binding
          ubiquitin protein ligase binding
          proton-transporting ATP synthase activity, rotational mechanism
          proton-transporting ATPase activity, rotational mechanism

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline