Search
Search

Entrez Gene ID: | 63891 |
Gene Name: | ring finger protein 123 |
Gene Aliases: |
FP1477, KPC1 |
Location: |
Chr.3:49688753-49721529 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF123 | NM_022064.4 | NP_071347.2 | ||
| NR_135218.1 | ||||
| XM_006713285.1 | XP_006713348.1 | |||
| XM_011533995.1 | XP_011532297.1 | |||
| XM_017007018.1 | XP_016862507.1 | |||
| AK096353.1 | ||||
| AK126883.1 | ||||
| AK308130.1 | ||||
| AK308607.1 | ||||
| AL832034.1 | ||||
| AL832138.1 | ||||
| AL832344.1 | ||||
| BC130632.1 | ||||
| BI596589.1 | ||||
| CR627051.1 | ||||
| DA334003.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1148681 | Chr.3:49683166 - 49692167 on Build GRCh38 | Duplication |
|
| nsv1118699 | Chr.3:49686367 - 49691867 on Build GRCh38 | Duplication |
|
| nsv1141429 | Chr.3:49676267 - 49693067 on Build GRCh38 | Duplication |
|
| esv28815 | Chr.3:49683318 - 49698418 on Build GRCh38 | Gain+Loss |
|
| nsv822077 | Chr.3:49683967 - 49697093 on Build GRCh38 | Gain |
|
| nsv508217 | Chr.3:49578013 - 49732463 on Build GRCh38 | Deletion |
|
| nsv820990 | Chr.3:49683218 - 49698478 on Build GRCh38 | Duplication |
|
Set Membership: |
|