Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm00186822_cn
See other GAPDH CNV Assays ›
Gene Symbol
Gapdh
Assay Reference Genome
Location
Chr.6:125165285 on build GRCm38
Cytoband
6F2
Assay ID
Mm00186822_cn
Size
S: 360 rxns
M: 750 rxns
L: 2,900 rxns
Availability
Made To Order
Catalog #
4400291
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Target Gene Details
Entrez Gene ID:
14433
Gene Name:
glyceraldehyde-3-phosphate dehydrogenase
Gene Aliases:
Gapd
Location:
Chr.6:125161338-125166511 on Build GRCm38
Assay Gene Location:
Overlaps Intron 2 - Intron 3
Gene Symbol
Transcript Accession
Exon Location
Assay Transcript Location
Protein ID
Gapdh
NM_001289726.1
NP_001276655.1
NM_008084.3
NP_032110.1
AK002273.1
BAB21979.1
AK081405.1
BAC38211.1
AK140794.1
BAE24481.1
AK144690.1
BAE26016.1
AK146435.1
BAE27169.1
AK147738.1
BAE28105.1
AK147891.1
BAE28208.1
AK160399.1
BAE35768.1
AK160753.1
BAE35989.1
AK164415.1
BAE37778.1
AK168217.1
BAE40174.1
AK184442.1
AK189223.1
AK190771.1
AK191938.1
AK192040.1
AK198590.1
AK199549.1
AK199608.1
AK201736.1
BC082592.1
AAH82592.1
BC083065.1
AAH83065.1
BC083079.1
AAH83079.1
BC083080.1
AAH83080.1
BC085275.1
AAH85275.1
BC085315.1
AAH85315.1
BC092252.1
AAH92252.1
BC095932.1
AAH95932.1
BC096440.1
AAH96440.1
BC110311.1
AAI10312.1
BC145810.1
BC145812.1
CJ155387.1
GU214026.1
M32599.1
AAA37659.1
Back To Top
More Information
Set Membership:
Intragenic
Intronic
Non-exonic
Panther Classification:
Molecular Function -
dehydrogenase
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
carbohydrate metabolic process
gluconeogenesis
glycolytic process
apoptotic process
multicellular organism development
negative regulation of translation
peptidyl-cysteine S-trans-nitrosylation
protein stabilization
neuron apoptotic process
oxidation-reduction process
response to ammonium ion
cellular response to interferon-gamma
glyceraldehyde-3-phosphate dehydrogenase (NAD+) (phosphorylating) activity
microtubule binding
peptidyl-cysteine S-nitrosylase activity
identical protein binding
Back To Top
Related Products
TaqMan
®
Genotyping Master Mix
TaqPath
®
ProAmp
®
Master Mix
TaqMan
®
Universal Master Mix II, no UNG
TaqMan
®
Gene Expression Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Universal PCR Master Mix, No AmpErase UNG
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline