Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00186825_cn
          See other GAPDH CNV Assays ›
          Gene Symbol
          Gapdh
          Assay Reference Genome
          Location

          Chr.6:125162863 on build GRCm38
          Cytoband
          6F2
          Assay ID Mm00186825_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          14433

          Gene Name:

          glyceraldehyde-3-phosphate dehydrogenase

          Gene Aliases:

          Gapd

          Location:

          Chr.6:125161338-125166511 on Build GRCm38

          Assay Gene Location:

          Overlaps Intron 5 - Exon 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          Gapdh NM_001289726.1 NP_001276655.1
          NM_008084.3 NP_032110.1
          AK002273.1 BAB21979.1
          AK081405.1 BAC38211.1
          AK140794.1 BAE24481.1
          AK144690.1 BAE26016.1
          AK146435.1 BAE27169.1
          AK147738.1 BAE28105.1
          AK147891.1 BAE28208.1
          AK160399.1 BAE35768.1
          AK160753.1 BAE35989.1
          AK164415.1 BAE37778.1
          AK168217.1 BAE40174.1
          AK169742.1 1 2142
          AK178680.1
          AK184044.1
          AK184664.1
          AK184690.1
          AK185135.1
          AK193664.1
          AK197268.1
          AK200636.1
          AK201486.1
          AK204478.1
          AK213421.1
          AK218066.1
          AK218240.1
          BC082592.1 AAH82592.1
          BC083065.1 AAH83065.1
          BC083079.1 AAH83079.1
          BC083080.1 AAH83080.1
          BC085275.1 AAH85275.1
          BC085315.1 AAH85315.1
          BC092252.1 AAH92252.1
          BC095932.1 AAH95932.1
          BC096440.1 AAH96440.1
          BC110311.1 AAI10312.1
          BC145810.1
          BC145812.1
          BI851476.1
          DQ403054.1
          GU214026.1
          M32599.1 AAA37659.1

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          carbohydrate metabolic process
          gluconeogenesis
          glycolytic process
          apoptotic process
          multicellular organism development
          negative regulation of translation
          peptidyl-cysteine S-trans-nitrosylation
          protein stabilization
          neuron apoptotic process
          oxidation-reduction process
          response to ammonium ion
          cellular response to interferon-gamma
          glyceraldehyde-3-phosphate dehydrogenase (NAD+) (phosphorylating) activity
          microtubule binding
          peptidyl-cysteine S-nitrosylase activity
          identical protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline