Search
Search

Entrez Gene ID: | 11831 |
Gene Name: | aquaporin 6 |
Gene Aliases: |
AI790337 |
Location: |
Chr.15:99599053-99605477 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aqp6 | NM_175087.4 | NP_780296.1 | ||
| AK082699.1 | BAC38576.1 | |||
| BC115586.1 | 4 | 821 | ||
| BC115587.1 | 4 | 820 | ||
| DQ464060.1 | 4 | 821 | ||
| DQ826418.1 |
Set Membership: |
|