Search
Search

Entrez Gene ID: | 73458 |
Gene Name: | aldehyde dehydrogenase 3 family, member B3 |
Gene Aliases: |
1700055N04Rik |
Location: |
Chr.19:3958733-3972419 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh3b3 | NM_028545.2 | NP_082821.2 | ||
| AK006803.1 | BAB24748.1 | |||
| AK081788.1 |
Set Membership: |
|