Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mr00733719_cn
See other ICRE CNV Assays ›
Gene Symbol
iCre
Assay Reference Genome
Location
-
Cytoband
-
Assay ID
Mr00733719_cn
Size
S: 360 rxns
M: 750 rxns
L: 2,900 rxns
Availability
Made To Order
Catalog #
4400291
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Target Gene Details
Gene Symbol:
iCre
Gene Name:
Synthetic construct: codon-optimized cre recombinase (iCre) gene
Gene Aliases:
-
Back To Top
More Information
Important Information
Mr00733719_cn is designed to detect codon-optimized cre recombinase (iCre) gene sequences used in some mammalian transgene vectors for optimized expression. A reference sequence for the assay is GeneBank AY056050.1.
Back To Top
Related Products
TaqMan
®
Genotyping Master Mix
TaqPath
®
ProAmp
®
Master Mix
TaqMan
®
Universal Master Mix II, no UNG
TaqMan
®
Gene Expression Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Universal PCR Master Mix, No AmpErase UNG
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline