Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other COL1A1 EBGEX Assays ›
          Gene Symbol:
          COL1A1
          Gene Name:
          collagen type I alpha 1
          Gene Aliases:
          EDSC, OI1, OI2, OI3, OI4
          Chromosome Location:
          Chr.17: 50184096 - 50201648 on Build GRCh38
          UniGene ID:
          Hs.172928
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          ProductType:
          QuantiGene Singleplex RNA
          Assay ID SA-10336
          Size
          Availability Made To Order
          Catalog # QGS-200
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: COL1A1
          Interrogated Sequence Translated Protein IMAGE Clone ID
          RefSeq NM_000088.3 NP_000079.2
          XM_005257058.3 XP_005257115.2
          XM_005257059.4 XP_005257116.2
          XM_011524341.1 XP_011522643.1
          GenBank mRNA AB209597.1 -
          AK297731.1 -
          BC036531.2 - 5264710
          DQ590066.1 -
          ENST00000225964.0 -
          JQ236861.1 -
          K01228.1 -
          M32798.1 -
          M36546.1 -
          S64596.1 -
          S64717.1 -
          S67495.1 -
          X06269.1 -
          X07884.1 -
          Z74615.1 -

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          blood vessel development
          osteoblast differentiation
          intramembranous ossification
          endochondral ossification
          blood coagulation
          visual perception
          sensory perception of sound
          positive regulation of epithelial to mesenchymal transition
          negative regulation of cell-substrate adhesion
          protein transport
          platelet activation
          extracellular matrix organization
          collagen fibril organization
          positive regulation of cell migration
          collagen catabolic process
          response to corticosteroid
          response to estradiol
          collagen biosynthetic process
          protein localization to nucleus
          tooth mineralization
          response to drug
          response to hydrogen peroxide
          response to peptide hormone
          skin morphogenesis
          cellular response to fibroblast growth factor stimulus
          tooth eruption
          positive regulation of transcription, DNA-templated
          embryonic skeletal system development
          regulation of immune response
          leukocyte migration
          response to cAMP
          response to hyperoxia
          face morphogenesis
          bone trabecula formation
          cartilage development involved in endochondral bone morphogenesis
          protein heterotrimerization
          cellular response to amino acid stimulus
          cellular response to mechanical stimulus
          cellular response to retinoic acid
          cellular response to vitamin E
          cellular response to tumor necrosis factor
          cellular response to epidermal growth factor stimulus
          cellular response to transforming growth factor beta stimulus
          positive regulation of canonical Wnt signaling pathway
          cellular response to fluoride
          extracellular matrix structural constituent
          protein binding
          identical protein binding
          metal ion binding
          platelet-derived growth factor binding

          Back To Top

          Related Products

          • QuantiGene Singleplex Assay Kit
          • QuantiGene Singleplex HT Assay Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0