Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Morc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_177719.4 | NP_808387.2 | ||||
GenBank mRNA | AK077016.1 | - | ||||
BC120896.1 | - | 40061539 | ||||
BC120897.1 | - | 40061540 |