Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC104260.1 | - | 4 - 5 | 471 | 40039813 | 74 |
BC104261.1 | - | 4 - 5 | 471 | 40039814 | 74 | |
DQ358978.1 | - | 4 - 5 | 410 | 74 | ||
U08030.1 | - | 4 - 5 | 469 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |