Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnpepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181405.4 | NP_852070.3 | 6 - 7 | 1416 | 70 | |
GenBank mRNA | CX218581.1 | - | 2 - 3 | 302 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |