Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266270.1 | NP_001253199.1 | 656 | 60 | ||
XM_015135403.2 | XP_014990889.1 | 684 | 60 | |||
GenBank mRNA | JU475493.1 | - | 656 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |