Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aaas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106795.1 | NP_001100265.1 | 5 - 6 | 627 | 118 | |
XM_006242387.2 | XP_006242449.1 | 4 - 5 | 544 | 118 | ||
XM_008765704.1 | XP_008763926.1 | 4 - 5 | 587 | 118 | ||
XM_008765705.1 | XP_008763927.1 | 4 - 5 | 568 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |