Search
Search

ABCD1| Gene Symbol: ABCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204043.2 | NP_001190972.1 | 24 - 25 | 4309 | 72 | |
| NM_001204044.1 | NP_001190973.1 | 24 - 25 | 4264 | 72 | ||
| NM_001342558.1 | NP_001328232.1 | 23 - 24 | 4628 | 72 | ||
| NM_120148.3 | NP_568072.1 | 24 - 25 | 4251 | 72 | ||
| GenBank mRNA | AF378120.1 | - | 24 - 25 | 4180 | 72 | |
Set Membership: |
|