Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339858.1 | NP_001319779.1 | 11 - 12 | 1398 | 71 | |
NM_001339859.1 | NP_001325644.1 | 11 - 12 | 1398 | 71 | ||
NM_001339860.1 | NP_001325645.1 | 11 - 12 | 2163 | 71 | ||
NM_001339861.1 | NP_001325651.1 | 11 - 12 | 2217 | 71 | ||
NM_001339862.1 | NP_001325648.1 | 11 - 12 | 1832 | 71 | ||
NM_001339863.1 | NP_001325647.1 | 11 - 12 | 1870 | 71 | ||
NM_001339864.1 | NP_001325650.1 | 11 - 12 | 1378 | 71 | ||
NM_001339865.1 | NP_001325649.1 | 10 - 11 | 1534 | 71 | ||
NM_001339866.1 | NP_001325646.1 | 10 - 11 | 1572 | 71 | ||
NM_115601.3 | NP_567048.1 | 11 - 12 | 1436 | 71 | ||
GenBank mRNA | AK318626.1 | - | 10 - 11 | 1389 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |