Search
Search

PSRP6| Gene Symbol: PSRP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343528.1 | NP_001331057.1 | 1 | 676 | 79 | |
| NM_121793.3 | NP_568358.1 | 1 | 490 | 79 | ||
| GenBank mRNA | AY086669.1 | - | 1 | 484 | 79 | |
Set Membership: |
|