Search
Search

ALDH12A1| Gene Symbol: ALDH12A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125647.4 | NP_568955.1 | 13 - 14 | 1487 | 97 | |
| GenBank mRNA | AY065072.1 | - | 13 - 14 | 1384 | 97 | |
| BT000746.1 | - | 13 - 14 | 1417 | 97 | ||
| BT010391.1 | - | 13 - 14 | 1334 | 97 | ||
Set Membership: |
|