Search
Search

MIPS3| Gene Symbol: MIPS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121055.4 | NP_196579.1 | 5 - 6 | 1028 | 110 | |
| GenBank mRNA | AY065415.1 | - | 5 - 6 | 958 | 110 | |
| AY096554.1 | - | 5 - 6 | 868 | 110 | ||
Set Membership: |
|