Search
Search

CYP82F1| Gene Symbol: CYP82F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335975.1 | NP_001325244.1 | 1 - 2 | 1000 | 87 | |
| NM_128073.3 | NP_180088.1 | 1 - 2 | 1000 | 87 | ||
| GenBank mRNA | AK229524.1 | - | 1 - 2 | 959 | 87 | |
| BT008597.1 | - | 1 - 2 | 959 | 87 | ||
| BT014968.1 | - | 1 - 2 | 930 | 87 | ||
Set Membership: |
|