Search
Search

CEACAM1| Gene Symbol: CEACAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205788.1 | NP_991357.1 | 1 - 2 | 101 | 78 | |
| GenBank mRNA | AY345127.1 | - | 1 - 2 | 101 | 78 | |
| AY345128.1 | - | 1 - 2 | 101 | 78 | ||
| AY345129.1 | - | 1 - 2 | 101 | 78 | ||
| AY345130.1 | - | 1 - 2 | 101 | 78 | ||
| AY487416.1 | - | 1 - 2 | 101 | 78 | ||
| AY487417.1 | - | 1 - 2 | 101 | 78 | ||
| AY487418.1 | - | 1 - 2 | 101 | 78 | ||
| AY487419.1 | - | 1 - 2 | 101 | 78 | ||
| BC120204.1 | - | 1 - 2 | 164 | 78 | ||
Set Membership: |
|