Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502334.1 | XP_003502382.1 | 1 - 2 | 101 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |