Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GlcAT-P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259784.2 | NP_001246713.1 | 4 - 5 | 947 | 103 | |
NM_140191.3 | NP_648448.1 | 4 - 5 | 1048 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |