Search
Search

sowi| Gene Symbol: sowi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141414.2 | NP_649671.2 | 1 - 2 | 506 | 84 | |
| GenBank mRNA | BT022649.1 | - | 1 - 2 | 505 | 84 | |
| BT022678.1 | - | 1 - 2 | 508 | 84 | ||
Set Membership: |
|