Search
Search

Ada2a| Gene Symbol: Ada2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014634.3 | NP_001014634.1 | 2 - 3 | 1036 | 70 | |
| NM_001014635.3 | NP_001014635.1 | 2 - 3 | 991 | 70 | ||
| NM_001014636.3 | NP_001014636.1 | 2 - 3 | 1036 | 70 | ||
| NM_001014637.3 | NP_001014637.1 | 2 - 3 | 991 | 70 | ||
| NM_001170174.2 | NP_001163645.1 | 2 - 3 | 991 | 70 | ||
| GenBank mRNA | BT025205.1 | - | 2 - 3 | 962 | 70 | |
Set Membership: |
|