Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ankrd29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020760.1 | NP_001018596.1 | 3 - 4 | 491 | 65 | |
XM_005161708.4 | XP_005161765.1 | 3 - 4 | 352 | 65 | ||
GenBank mRNA | BC095639.1 | - | 3 - 4 | 491 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |