| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000429938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000429938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000429938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000429938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000429938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006761.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006761.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006761.hg.1 |
TC07000485.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006766.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006766.hg.1 |
ENST00000432276.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006766.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006766.hg.1 |
TC07000485.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006767.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006767.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006767.hg.1 |
TC07000485.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006762.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006762.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006762.hg.1 |
TC07000485.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006772.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006772.hg.1 |
ENST00000429938.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006772.hg.1 |
ENST00000432276.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006772.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006772.hg.1 |
TC07000485.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006760.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006760.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006760.hg.1 |
TC07000485.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010395.hg.1 |
ENST00000423240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010395.hg.1 |
ENST00000423240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010395.hg.1 |
ENST00000423240.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006768.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006768.hg.1 |
ENST00000432276.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006768.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07006768.hg.1 |
TC07000485.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009665.hg.1 |
ENST00000448722.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009665.hg.1 |
ENST00000448722.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156089.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156089.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156089.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156089.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156089.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156089.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156089.hg.1 |
TC0700008090.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000429938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000429938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000429938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000429938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000429938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008090.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010395.hg.1 |
ENST00000423240.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010395.hg.1 |
ENST00000423240.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010395.hg.1 |
ENST00000423240.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156083.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156083.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156083.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156083.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156083.hg.1 |
TC0700008090.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700156085.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156085.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156085.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156085.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156085.hg.1 |
TC0700008090.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700156084.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156084.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156084.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156084.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156084.hg.1 |
TC0700008090.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700156093.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156093.hg.1 |
ENST00000429938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156093.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156093.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156093.hg.1 |
ENST00000248553.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156093.hg.1 |
ENST00000429938.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156093.hg.1 |
ENST00000447574.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700156093.hg.1 |
NM_001540.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700156093.hg.1 |
TC0700008090.hg.1 |
Clariom D TC |