| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011106.hg.1 |
ENST00000427833.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011106.hg.1 |
ENST00000427833.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011106.hg.1 |
ENST00000427833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011106.hg.1 |
ENST00000427833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011106.hg.1 |
ENST00000427833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011106.hg.1 |
ENST00000427833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000395787.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
ENST00000527830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100013163.hg.1 |
NM_015957.3 |
Refseq |